Skip to Content
Merck
All Photos(1)

Key Documents

04-478

Sigma-Aldrich

Anti-phospho-STAT1 (Ser 727) Antibody, clone 12C5

clone 12C5, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

12C5, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer727)

Gene Information

human ... STAT1(6772)

Specificity

Recognizes phosphorylated domain of STAT1 on Serine727

Immunogen

KLH-conjugated synthetic peptide encompassing the surrounding amino acids of Serine727 on phosphorylated STAT1

Application

Anti-phospho-STAT1 (Ser 727) Antibody, clone 12C5 is a Mouse Monoclonal Antibody for detection of phospho-STAT1 (Ser 727) also known as signal transducer & activator of transcription 1 & has been tested in ELISA & WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

Routinely evaluated by immunoblot.

Target description

92 kDa

Physical form

Format: Purified
In 1 mL of PBS containing 0.09% sodium azide, PEG, sucrose, and 50% glycerol.
Subsequent thiophilic adsorption and size exclusion chromatography

Storage and Stability

2 years at -20°C from date of shipment. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

VEGF receptors on chronic lymphocytic leukemia (CLL) B cells interact with STAT 1 and 3: implication for apoptosis resistance.
Lee, Y K, et al.
Leukemia, 19, 513-523 (2005)
Rico K H Lo et al.
Cellular signalling, 18(3), 307-317 (2005-06-28)
The ability of the human prostacyclin receptor (hIP) to regulate the activities of signal transducers and activators of transcription (STATs) has not yet been documented. In the present study, we have delineated the mechanism by which hIP induces STAT3 phosphorylations
H Nguyen et al.
The Journal of biological chemistry, 276(36), 33361-33368 (2001-07-05)
STAT1 must be phosphorylated on serine 727 to be fully active in transcription. We show that phosphatidylinositol 3-kinase (PI3K) and its effector kinase Akt play an important role in the serine phosphorylation of STAT1 and in the activation of gene
Christopher P Gayer et al.
The Journal of biological chemistry, 284(4), 2001-2011 (2008-12-03)
The intestinal epithelium is repetitively deformed by shear, peristalsis, and villous motility. Such repetitive deformation stimulates the proliferation of intestinal epithelial cells on collagen or laminin substrates via ERK, but the upstream mediators of this effect are poorly understood. We
Rama S Kota et al.
The Journal of biological chemistry, 280(35), 31076-31084 (2005-07-05)
Lipoprotein lipase (LPL) is a key enzyme in the hydrolysis of triglyceride-rich lipoproteins. In vascular diseases, such as atherosclerosis, inflammation plays an important role in the pathogenesis of the disease. We examined the role of LPL in modulating tumor necrosis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service