Skip to Content
Merck
All Photos(1)

Documents

04-299

Sigma-Aldrich

Anti-phospho-Insulin Receptor (Tyr 1150/1151) Antibody, clone 10C3

clone 10C3, Upstate®, from mouse

Synonym(s):

INSR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

10C3, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pTyr1150/pTyr1151)

Gene Information

human ... INSR(3643)

Specificity

Recognizes phosphorylated domain of Insulin receptor on Tyrosine 1150/1151.

Immunogen

KLH-conjugated synthetic peptide encompassing the surrounding amino acids of Tyrosine 1150/1151 on phosphorylated insulin receptor.

Application

Detect phospho-Insulin Receptor (Tyr 1150/1151) using this Anti-phospho-Insulin Receptor (Tyr 1150/1151) Antibody, clone 10C3 validated for use in EA & WB.

Quality

Routinely evaluated by immunoblot.

Target description

97 kDa

Physical form

Format: Purified
In 1 mL of PBS containing 0.09% sodium azide, PEG, and sucrose and 50% glycerol.

Analysis Note

Control
Includes pervanadate treated HEK293 lysate as a positive control

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natalie N Rizk et al.
Fertility and sterility, 86(4 Suppl), 1217-1222 (2006-09-12)
To determine whether the insulin receptor (IR), insulin-like growth factor-I receptor (IGF-IR), and IGF-I are expressed differentially in fibroblasts isolated from normal peritoneal and adhesion tissue before and after 24-hour treatment with increasing glucose concentrations. Prospective experimental study. University medical
Rita Slaaby et al.
The Journal of biological chemistry, 281(36), 25869-25874 (2006-07-13)
Insulin receptor (IR) and insulin-like growth factor I receptor (IGF-IR) are both from the same subgroup of receptor tyrosine kinases that exist as covalently bound receptor dimers at the cell surface. For both IR and IGF-IR, the most described forms
Qiu-yan Wang et al.
The FEBS journal, 274(2), 526-538 (2007-01-19)
Two H7721 human hepatocarcinoma cell lines showing moderate and high expression of alpha1,3-fucosyltransferase (FucT)-VII cDNA were established and designated FucTVII-M and FucTVII-H, respectively. In alpha1,3-FucT-VII-transfected cells, expression of insulin receptor (InR) alpha- and beta subunits and epidermal growth factor receptor
Amer Youssef et al.
Endocrinology, 157(3), 1163-1174 (2016-01-14)
Placental mesenchymal stem cells (PMSCs) are readily available multipotent stem cells for potential use in regenerative therapies. For this purpose, PMSCs must be maintained in culture conditions that mimic the in vivo microenvironment. IGFs (IGF-1 and IGF-2) and oxygen tension
Atsumi Tsuji-Hosokawa et al.
Pediatric diabetes, 18(8), 917-924 (2017-02-10)
Defects of the insulin receptor gene ( INSR ) cause wide spectra of congenital insulin resistance. Monoallelic defects result in milder insulin-resistant diabetes mellitus with acanthosis nigricans (IRAN, type A). Whereas, leprechaunism (Donahue syndrome), the most severe condition with lethality

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service