Skip to Content
Merck
All Photos(2)

Documents

03-248

Sigma-Aldrich

RIPAb+ pan Ago - RIP Validated Antibody and Primer Set

clone 2A8, from mouse

Synonym(s):

Eukaryotic translation initiation factor 2C, 2, Protein argonaute-2, Ago2, Protein slicer, Argonaute2, PAZ Piwi domain protein, Ago4, Argonaute4, Protein argonaute-4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.32

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

clone

2A8, monoclonal

species reactivity

human

manufacturer/tradename

RIPAb+
Upstate®

technique(s)

RIP: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

General description

RIPAb+ antibodies are evaluated using the RNA Binding Protein Immunoprecipitation (RIP) assay. Each RIPAb+ antibody set includes a negative control antibody to ensure specificity of the RIP reaction and is verified for the co-immunoprecipitation of RNA associated specifically with the immunoprecipitated RNA binding protein of interest. Where appropriate, the RIPAb+ set also includes quantitative RT-PCR control primers (RIP Primers) to biologically validate your IP results by successfully co-precipitating the specific RNA targets, such as messenger RNAs. The qPCR protocol and primer sequences are provided, allowing researchers to validate RIP protocols when using the antibody in their experimental context. If a target specific assay is not provided, the RIPAb+ kit is validated using an automated microfluidics-based assay by enrichment of detectable RNA over control immunoprecipitation.
Argonaute (Ago) proteins are a family of proteins of ~95 kDa that bind directly to mature miRNAs. In mammals, there are four Ago proteins (Ago1-4). This family of proteins is a central component of most mammalian miRNPs that mediates important functions in small RNA-directed regulatory pathways.

Specificity

This antibody recognizes Ago1, Ago2, Ago3, and Ago4. Note that this antibody has been reported to cross-react with radixin (~70 kDa). (Nelson, P., et al. (2007). RNA. 13:1787–1792.)

Immunogen

Histidine - tagged recombinant protein corresponding to human Ago.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

RNA Metabolism & Binding Proteins
This RIPAb+ pan Ago -RIP Validated Antibody & Primer Set conveniently includes the antibody & the specific control PCR primers.
Western Blot Analysis:
Representative lot data.
HeLa cell lysate was probed with Anti-pan Ago, clone 2A8 (0.5 μg/mL). Proteins were visualized using a Goat Anti-Mouse IgG secondary antibody conjugated to HRP and chemiluminescence detection system.
Arrows indicates Ago (~ 95 kDa) and Radixin (~70 kDa). (Figure 2).
Immunoprecipitation Analysis: A representative lot was used by an independent laboratory in IP. (Nelson, P., et al. (2007). RNA. 13:1787–1792.)
Immunocytochemistry Analysis: A representative lot was used by an independent laboratory in IC. (Nelson, P., et al. (2007). RNA. 13:1787–
1792.)
Immunohistochemistry Analysis: A representative lot was used by an independent laboratory in IH. (Nelson, P., et al. (2007). RNA. 13:1787–
1792.)

Packaging

10 assays per set. Recommended use: ~5 μg of antibody per RIP (dependent upon biological context).

Quality

RNA Binding Protein Immunoprecipitation:
Representative lot data.
RIP Lysate prepared from 293 cells (2 X 10E7 cell equivalents per IP) were subjected to immunoprecipitation using 5 µg of either a normal mouse IgG or 5 µg of Anti-pan Ago antibodyand the Magna RIP® RNA-Binding Protein Immunoprecipitation Kit (Cat. # 17-700).
Successful immunoprecipitation of Argonaute proteins-associated RNA was verified by qPCR using RIP Primers IGF2, (Figure 1).
Please refer to the Magna RIP (Cat. # 17-700) or EZ-Magna RIP (Cat. # 17-701) protocol for experimental details.

Target description

~ 95 and 70 kDa observed. This antibody recognizes Ago1, Ago2, Ago3, and Ago4.
Note that this antibody has been reported to cross-react with radixin (~70 kDa). (Nelson, P., et al. (2007). RNA. 13:1787–1792.)

Physical form

Anti-pan Ago (Mouse Monoclonal).One vial containing 50 μg of purified mouse IgG1κ in 0.1 M Tris-Glycine (pH 7.4) and 150 mM NaCl with 0.05% sodium azide before the addition of glycerol to 30%. Store at -20°C.
Normal Mouse IgG. One vial containing 125 µg of purified mouse IgG in 125 µL of storage buffer containing 0.1% sodium azide. Store at -20°C.
RIP Primer IGF2. One vial containing 75 μL of 5 μM of each primer specific for human IGF2 mRNA. Store at -20°C.
FOR: GCG GCT TCT ACT TCA GCA G
REV: CAG GTG TCA TAT TGG AAG AAC
Format: Purified
Protein G

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. Note: Variabillity in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Analysis Note

Control
Includes negative control normal mouse IgG antibody and control primers specific for the cDNA of human IGF2 mRNA.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

MAGNA RIP is a registered trademark of Merck KGaA, Darmstadt, Germany
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A high throughput experimental approach to identify miRNA targets in human cells.
Tan, Lu Ping, et al.
Nucleic Acids Research, 37, e137-e137 (2009)
Nannan Zhang et al.
Current protocols in immunology, 126(1), e78-e78 (2019-09-05)
MicroRNAs (miRNAs) are short (19- to 25-nucleotide) noncoding RNA molecules that target mRNAs to repress gene expression and that play important roles in regulating many fundamental biological functions including cell differentiation, development, growth, and metabolism. They are well conserved in
Qiang Guan et al.
Experimental and therapeutic medicine, 22(1), 755-755 (2021-05-27)
Hepatocellular carcinoma (HCC) is the third leading cause of cancer-related death worldwide with a low 5-year survival rate. Long non-coding RNA (lncRNA) double homeobox A pseudogene 8 (DUXAP8) is an oncogene and a potential biomarker in various tumors, such as
Tanner C Godfrey et al.
The Journal of biological chemistry, 293(45), 17646-17660 (2018-09-23)
MicroRNAs (miRs) and Hox transcription factors have decisive roles in postnatal bone formation and homeostasis. In silico analysis identified extensive interaction between HOXA cluster mRNA and microRNAs from the miR-23a cluster. However, Hox regulation by the miR-23a cluster during osteoblast
Mathieu Bertrand et al.
Developmental dynamics : an official publication of the American Association of Anatomists, 230(2), 325-334 (2004-05-27)
The MAGED gene subfamily contains three genes in mouse and four in human. The MAGED1, D2, and D3 proteins are highly conserved between mouse and human, whereas paralogues are less conserved between each other. This finding suggests that each MAGED

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service