Skip to Content
Merck

Skip To

03-230

SIRT5 (His-tag) human recombinant - HDAC protein

Human recombinant SIRT5 (His-tag) expressed in E. Coli.

Synonym(s):

HDAC SIRT5, His-tagged SIRT5, SIRT5 Human Recombinant

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352200
NACRES:
NA.26
eCl@ss:
32160405
Assay:
>95% (SDS-PAGE)
Biological source:
Escherichia coli, human
Form:
liquid
Solubility:
H2O: soluble at 20 °C
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

biological source

Escherichia coli, human

recombinant

expressed in E. coli

assay

>95% (SDS-PAGE)

form

liquid

specific activity

>200 pmol/min-mg, 37 °C (fluorometric activity assay)

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable

solubility

H2O: soluble at 20 °C

shipped in

dry ice

General description

Human recombinant SIRT5 (His-tag) expressed in E. Coli
Product Source: Recombinant residues 37-310 of human SIRT5 isoform 1, with an N-terminal His-tag

Application

HDAC deacetylase assays
Human recombinant SIRT5 (His-tag) expressed in E. Coli.
Research Category
Epigenetics & Nuclear Function

Physical form

Format: Purified
In liquid containing 25 mM Tris, pH 7.5, 100 mM NaCl, 5 mM DTT, containing 10% glycerol. Frozen solution.

Preparation Note

Store at -80°C. The enzyme is stable on ice for 30 to 60 min, but may lose activity with dilution and/or prolonged storage on ice. It is recommended that thawing and addition of the enzyme to assay wells be done within as short a time as possible before start of the assay and that the thawed enzyme be kept on ice throughout. Best performance will be obtained by adding the chilled, undiluted enzyme directly to pre-warmed buffer and proceeding immediately to the addition of pre-warmed substrate. The remaining, unused enzyme should be refrozen quickly by, for example, snap freezing in a dry/ice ethanol bath or liquid nitrogen. The enzyme is stable to at least 4 freeze/thaw cycles. To minimize the number of freeze/thaw cycles, aliquot the SIRT5 into separate tubes before storing at -80°C.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB570059614-69714-865
biological source

Escherichia coli, human

biological source

-

biological source

human

biological source

human

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

manufacturer/tradename

Upstate®

manufacturer/tradename

-

manufacturer/tradename

Upstate®

manufacturer/tradename

Upstate®

form

liquid

form

liquid

form

-

form

liquid

assay

>95% (SDS-PAGE)

assay

>95% (HPLC), >95% (SDS-PAGE)

assay

-

assay

~95% (SDS-PAGE)

solubility

H2O: soluble at 20 °C

solubility

-

solubility

-

solubility

water: soluble


wgk

WGK 2

flash_point_f

does not flash

flash_point_c

does not flash



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
03-23004053252550911

Questions

Reviews

No rating value

Active Filters