Skip to Content
Merck
All Photos(1)

Key Documents

03-0204-00

Millipore

SMC® Human IgM High Sensitivity Immunoassay Kit

1 kit sufficient for 96 wells

Synonym(s):

IgM Single Molecule Counting Kit, IgM ultra-sensitive immunoassay, Immunoglobulin M immunoassay kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84
Pricing and availability is not currently available.

Quality Level

usage

 kit sufficient for 96 wells

species reactivity

human

manufacturer/tradename

SMC®

assay range

accuracy: 70-130%
(minimum of 5 samples spiked)

inter-assay cv: 12%
intra-assay cv: 7%

technique(s)

ELISA: suitable
single molecule counting: suitable

compatibility

for use with SMCxPRO® and Erenna (validated)

shipped in

wet ice

storage temp.

2-8°C

General description

The Single Molecule Counting (SMC® Human IgM High Sensitivity Immunoassay uses a quantitative fluorescent sandwich immunoassay technique to measure IgM in human serum and plasma samples. The user coats a capture antibody specific for human IgM onto a 96-well microplate. After blocking the plate, the user pipettes standards and samples into the coated microplate wells. During incubation, the IgM present in the sample binds to the capture antibody on the coated plate. Unbound molecules are washed away during a wash step. The fluor-labeled detection antibody is added to each well and incubated. The detection antibody recognizes and binds to IgM that has been captured onto the plate, thus completing the immunosanwich. Following the final wash step, elution buffer is added and incubated. The elution buffer dissociates the bound protein sandwich from the plate surface, releasing the labeled antibodies. These antibodies are separated during transfer to a final microplate. The reading plate is loaded onto the Erenna or SMCxPRO® System where the labeled molecules are detected and counted. The number of fluor-labeled detection antibodies counted is directly proportional to the amount of IgM present in the sample when captured. The amount of IgM in unknown samples is interpolated from a standard curve.

The SMC® Human IgM High Sensitivity Immunoassay Kit contains all reagents required to perform the assays. This assay takes advantage of a standard immunoassay workflow configured in a standard plate format. Assay protocols are similar to existing sandwich ELISA methods with two key differences: 1) Elution buffer disrupts the sandwich, separating the labeled detection antibody for quantification. 2) The SMC®immunoassay system detects analytes using Single Molecule Counting (SMC®) technology.

Specificity

This assay does not show cross-reactivity against immunoglobulin subclasses IgA, IgD, IdE, IgG1, IgG2, IgG3, or IgG4 from in-house testing

Application

The Single Molecule Counting (SMC®) technology gives the SMC® Human IgM High Sensitivity Immunoassay Kit offers researchers the ability to successfully overcome sample matrix challenges when accurately quantifying human IgM in serum and plasma samples

Packaging

96 well assay

Components

  • Assay Plate
  • Coating Buffer
  • Blocking Buffer
  • Standard Diluent
  • IgM Detection Antibody
  • IgM Standard
  • SMC® Human IgM Immunoassay Kit Instructions
  • 10X Wash Buffer
  • Buffer C
  • Elution Buffer B
  • Buffer 3
  • IgM Coating Antibody

Legal Information

SMC is a registered trademark of Singulex
SMCxPRO is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Explore ultrasensitive immunoassay technology with SMC® technology, including its biomarker detection platform, advantages of high sensitivity assays, and more.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service