03-0159-00
SMC® Human IL-17A High Sensitivity Kit
1 kit sufficient for 96 wells, input: plasma, serum
Synonym(s):
IL-17A Single Molecule Counting Kit, IL-17A immunoassay kit, IL-17A ultra-sensitive immunoassay, Interleukin-17A immunoassay kit
About This Item
Recommended Products
usage
kit sufficient for 96 wells
species reactivity
human
manufacturer/tradename
SMC®
assay range
sensitivity: 0.07 pg/mL
(lower limit of quantification)
inter-assay cv: <15%
intra-assay cv: <10%
standard curve range: 0.02-60 pg/mL
technique(s)
ELISA: suitable
single molecule counting: suitable
input
plasma
serum
compatibility
for use with SMCxPRO® and Erenna<TMSYMBOL></TMSYMBOL> (validated)
shipped in
wet ice
storage temp.
2-8°C
General description
Assay protocols are similar to existing sandwich ELISA methods with two key differences:
1) Elution buffer disrupts the sandwich, separating the labeled detection antibody for quantification.
2)The Erenna and SMCxPRO® immunoassay systems detect analytes using Single Molecule Counting (SMC®) technology.
Specificity
Legal Information
Disclaimer
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 1B
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
To determine protein content, the Warburg-Christian method refers to measuring protein samples at 280 nm using a spectrophotometer.
Related Content
Protein quantification methods, reagents, and immunoassay technology for accurately measuring the protein concentrations in a variety of samples.
Explore ultrasensitive immunoassay technology with SMC® technology, including its biomarker detection platform, advantages of high sensitivity assays, and more.
Learn how immunoassays are used in cosmetics and personal care research to discover potential harmful effects of cosmetics products, and assess markers of inflammation, sensitization, aging, and tissue regeneration among others.
Use the power of high sensitivity immunology assays with SMC® technology to precisely characterize low-abundant cytokines like those from Th1/Th2 and Th17 cells.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service