Skip to Content
Merck
All Photos(1)

Key Documents

T-089

Supelco

2,5-Dimethoxy-4-(n)-propylthiophenethylamine-13C,D3 hydrochloride solution

100 μg/mL in acetonitrile: water (9:1) (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

2C-T-7-13C,D3 hydrochloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C1213CH18D3NO2S · HCl
Molecular Weight:
295.85
UNSPSC Code:
41116107
Pricing and availability is not currently available.

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule D (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIA (Portugal)

concentration

100 μg/mL in acetonitrile: water (9:1) (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

shipped in

wet ice

storage temp.

−20°C

General description

2C-T-7 is a psychedelic phenethylamine related to the 2C family of recreational drugs such as 2C-T-4, 2C-B, 2C-E, and 2C-I. Hallucinogenic effects often last from 8 to 15 hours after dosing. The designer drug is sold under street names including Blue Mystic and 7th Heaven. This stable-labeled internal standard is suitable for quantitation of 2C-T-7 levels in urine, serum, or plasma by GC/MS or LC/MS for use in clinical toxicology, forensic analysis, or urine drug testing applications.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service