Skip to Content
Merck
All Photos(1)

Key Documents

T-074

Supelco

3,3′,5-Triiodo-L-thyronine (T3) solution

100 μg/mL in methanol with 0.1N NH3, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

3,3′,5-Triiodo-L-thyronine, O-(4-Hydroxy-3-iodophenyl)-3,5-diiodo-L-tyrosine, Liothyronine, T3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12I3NO4
CAS Number:
Molecular Weight:
650.97
Beilstein:
2710227
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol with 0.1N NH3

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

mp

234-238 °C (lit.)

application(s)

clinical testing
clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

N[C@@H](Cc1cc(I)c(Oc2ccc(O)c(I)c2)c(I)c1)C(O)=O

InChI

1S/C15H12I3NO4/c16-9-6-8(1-2-13(9)20)23-14-10(17)3-7(4-11(14)18)5-12(19)15(21)22/h1-4,6,12,20H,5,19H2,(H,21,22)/t12-/m0/s1

InChI key

AUYYCJSJGJYCDS-LBPRGKRZSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Certified Spiking Solutions® are suitable for use as a starting material in the preparation of linearity standards, calibrators, and controls for thyroid hormone testing methods by LC-MS/MS.

3,3′,5-Triiodo-L-thyronine (T3) is a thyroid hormone and active form of L-thyroxine (T4), formed by deiodinase type I or II. It supports various physiological functions, such as oxygen consumption, carbohydrate metabolism, protein synthesis, and neural development. Free and total T3 serum levels are measured by LC-MS/MS for the determination of hyperthyroidism caused by excess T3.

Application

The Certified Reference Material (CRM) in solution can also be used as follows:

  • Multi-residue determination of seven thyroid hormones in placenta samples by isotope dilution liquid chromatography-quadrupole time-of-flight mass spectrometry (LC-Q-TOF-MS)
  • Parts-per-trillion analysis of five thyroid hormones in zebrafish larvae by their solid phase extraction (SPE) and ultra-high performance liquid chromatography-electrospray ionization- tandem mass spectrometric (UPLC-ESI/MS/MS) analysis
  • Simultaneous analysis of six free thyroid hormones in bovine serum samples by ultrafiltration and high-performance liquid chromatography (HPLC)-tandem mass spectrometry (MS/MS)
  • Determination of six thyroid hormones in mouse tissue samples by isotope dilution nano-ultraperformance liquid chromatography–quadrupole time-of-flight mass spectrometry (Q-TOF-MS)
  • Identification and quantification of triiodothyronine and thyroxine in rat serum samples using liquid chromatography-tandem mass spectrometry (LC-MS/MS)

Features and Benefits

  • Fully characterized under ISO/IEC 17025 and ISO 17034 accreditation
  • Accompanied with a comprehensive Certificate of Analysis (CoA) with data on stability, homogeneity, accuracy of concentration, uncertainty, and traceability
  • Rigorously tested through real-time stability studies to ensure accuracy and shelf life
  • Gravimetrically prepared using qualified precision balances to ensure minimal uncertainty
  • Flame sealed under argon into ampoules for long-term shelf life
  • Offered in a convenient, DEA-exempt format to improve laboratory efficiency

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTIONS is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vasiliki Michopoulos et al.
Fertility and sterility, 99(7), 2084-2091 (2013-03-20)
To determine whether cognitive behavior therapy (CBT), which we had shown in a previous study to restore ovarian function in women with functional hypothalamic amenorrhea (FHA), could also ameliorate hypercortisolemia and improve other neuroendocrine and metabolic concomitants of in FHA.
Min Kyong Moon et al.
European journal of endocrinology, 168(6), 805-810 (2013-03-21)
Body adiposity, especially ectopic fat accumulation, has a range of metabolic and cardiovascular effects. This study aimed to investigate whether thyroid function is associated with various regional fat quantities in euthyroid subjects. A total of 100 euthyroid men (free triiodothyronine
Susana B Bravo et al.
The Journal of clinical endocrinology and metabolism, 98(6), 2431-2441 (2013-03-30)
Mechanisms of thyroid physiology and cancer are principally studied in follicular cell lines. However, human thyroid cancer lines were found to be heavily contaminated by other sources, and only one supposedly normal-thyroid cell line, immortalized with SV40 antigen, is available.
Pragya Sharma et al.
The Journal of biological chemistry, 288(23), 16321-16333 (2013-05-01)
Secretory phospholipase A2 group IIa (PLA2g2a) is associated with inflammation, hyperlipidemia, and atherogenesis. Transcription of the PLA2g2a gene is induced by multiple cytokines. Here, we report the surprising observation that thyroid hormone (T3) inhibited PLA2g2a gene expression in human and
H T Celik et al.
Endocrine regulations, 47(2), 57-64 (2013-05-07)
Interleukin-33 (IL-33), a 30 kDa cytokine, is a member of IL-1 family. It is considered to be an autoimmune biomarker associated with T helper 2 (Th 2) response. γ-interferon is also produced by T helper 1 (Th 1) cells to

Articles

Determination of free thyroid hormones (T3 & T4) in serum by LC-MS/MS following a streamlined 1-hour BioSPME sample preparation.

Protocols

Determination of the thyroid hormones T4, T3 and rT3 from plasma using an Ascentis Express RP-Amide cartridge to trap the analytes, and a Phenyl-Hexyl column to resolve them.

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. May I know its corresponding Antibody?

    1 answer
    1. This product is a Certified Reference Material intended for use as an identification, calibration, and quantification standard. Unfortunately, a corresponding antibody is not available.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service