Skip to Content
Merck
All Photos(1)

Documents

S-149

Supelco

Serotonin-beta-D-glucuronide solution

new

100 μg/mL (ACN:H2O(50:50)(v/v)), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H20N2O7
CAS Number:
Molecular Weight:
352.34
MDL number:

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL (ACN:H2O(50:50)(v/v))

format

single component solution

storage temp.

-10 to -25°C

SMILES string

NCCC1=CNC2=CC=C(O[C@H](O3)[C@H](O)[C@@H](O)[C@H](O)[C@H]3C(O)=O)C=C21

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

48.2 °F

Flash Point(C)

9 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service