Skip to Content
Merck
All Photos(1)

Key Documents

P-038

Supelco

(±)-Phenylpropanolamine hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

DL-Norephedrine hydrochloride, Norephedrine hydrochloride, (±)-Phenylpropanolamine hydrochloride, erythro-2-Amino-1-phenyl-1-propanol hydrochloride, DL-erythro-2-Amino-1-phenyl-1-propanol hydrochloride, DL-Phenylpropanolamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OH)CH(NH2)CH3 · HCl
CAS Number:
Molecular Weight:
187.67
Beilstein:
3914299
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

mp

194-196 °C (lit.)

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

Cl.C[C@@H](N)[C@@H](O)c1ccccc1

InChI

1S/C9H13NO.ClH/c1-7(10)9(11)8-5-3-2-4-6-8;/h2-7,9,11H,10H2,1H3;1H/t7-,9-;/m1./s1

InChI key

DYWNLSQWJMTVGJ-PRCZDLBKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phenylpropanolamine, also known as Norephedrine, is a stimulant, decongestant, and anorectic drug commonly used in many prescription and over-the-counter cold and cough formulations. The drug is often abused illicitly for its psychoactive and stimulant effects. This Certified Spiking Solution® is applicable for use in many LC-MS/MS or GC/MS applications from clinical chemistry and forensic analysis to urine drug testing.

Biochem/physiol Actions

Mixed-acting sympathomimetic amine.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carlos Castillo-Henkel et al.
Proceedings of the Western Pharmacology Society, 53, 33-36 (2010-01-01)
Here we contrast the vascular smooth muscle contractility produced by D-nor-pseudoephedrine, alone or combined with triiodothyronine, on the aorta and coronary vasculature of the rat. At high concentrations (greater than those found in normal therapeutic dosing) D-nor-pseudoephedrine contracted the aorta.
Hyeon-Kyu Lee et al.
The Journal of organic chemistry, 77(12), 5454-5460 (2012-05-25)
Each of the enantiomers of both norephedrine and norpseudoephedrine were stereoselectively prepared from the common, prochiral cyclic sulfamidate imine of racemic 1-hydroxy-1-phenyl-propan-2-one by employing asymmetric transfer hydrogenation (ATH) catalyzed by the well-defined chiral Rh-complexes, (S,S)- or (R,R)-Cp*RhCl(TsDPEN), and HCO(2)H/Et(3)N as
John S Chappell et al.
Forensic science international, 195(1-3), 108-120 (2009-12-29)
A primary concern with the forensic analysis of the khat plant (Catha edulis) has been the need to preserve the principle psychoactive component, cathinone, which converts to the less-active substance, cathine, after harvesting. The loss of cathinone has serious legal
Saba Kassim et al.
Journal of ethnopharmacology, 140(1), 193-196 (2012-01-17)
Khat chewing amongst the UK communities originating from Yemen and the East African coast is suggested to create dependency through its main stimulant components (cathinone, norephedrine and norpseudoephedrine) on the central nervous system. To validate self-reported khat chewing behaviours by
A Pokrywka et al.
International journal of sports medicine, 30(8), 569-572 (2009-04-22)
Pseudoephedrine (PSE) as a sympathomimetic is an ingredient of many proprietary medicines which are available on the medical market over the counter (OTC drugs). It can be converted to cathine (CATH, norpseudoephedrine) inside the body. Until the end of 2003

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service