Skip to Content
Merck
All Photos(1)

Key Documents

P-023

Supelco

Phentermine solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15N
CAS Number:
Molecular Weight:
149.23
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIB (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

2-8°C

SMILES string

NC(C)(C)CC1=CC=CC=C1

InChI

1S/C10H15N/c1-10(2,11)8-9-6-4-3-5-7-9/h3-7H,8,11H2,1-2H3

InChI key

DHHVAGZRUROJKS-UHFFFAOYSA-N

General description

A Snap-N-Spike® reference solution applicable for use in clinical toxicology or urine drug testing by LC-MS/MS or GC/MS. Phentermine, a phenethylamine analog, is a stimulant and an appetite suppressant. This drug is a constituent of the withdrawn anti-obesity medication fen-phen.

Application

  • Phentermine solution for weight loss research: Phentermine is extensively studied as an appetite suppressant in obesity treatment. Its effectiveness in weight loss regimens is often analyzed through pharmaceutical research, where the solution form facilitates precise dosing and control in clinical trials, enhancing the understanding of its pharmacodynamics and therapeutic potential (Ni et al., 2015).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eva Manakova et al.
Neuro endocrinology letters, 33 Suppl 3, 179-182 (2013-01-29)
Both, obesity as well as anorexia may be associated with infertility and other complications of pregnancy. Weight loss during pregnancy is therefore considered a risk factor. Weight loss and appetite suppressant are contraindicated during pregnancy, but the unintended exposure is
Prescrire international, 22(136), 61-64 (2013-04-18)
The cornerstones of treatment for obesity, and even more so for simple overweight, are dietary measures and physical exercise. There are no drugs with a favourable harm-benefit balance in this setting. A fixed-dose combination of topiramate, an antiepileptic drug, and
Two new drugs approved for obesity.
Annette M Johnson
South Dakota medicine : the journal of the South Dakota State Medical Association, 65(9), 356-357 (2012-10-17)
David H Winslow et al.
Sleep, 35(11), 1529-1539 (2012-11-02)
To evaluate safety and efficacy of phentermine 15 mg plus extended-release topiramate 92 mg for treatment of moderate to severe obstructive sleep apnea (OSA) in obese adults. This phase 2, randomized, double-blind, placebo-controlled study included 2-week screening and 28-week treatment
Michael H Davidson et al.
The American journal of cardiology, 111(8), 1131-1138 (2013-02-05)
The aim of this analysis was to evaluate changes in cardiovascular risk factors in obese patients with dyslipidemia and/or hypertension receiving phentermine (PHEN) and topiramate extended-release (TPM ER). In the 56-week, randomized, double-blind, placebo-controlled, multicenter CONQUER trial, PHEN/TPM ER demonstrated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service