Skip to Content
Merck
All Photos(1)

Documents

O-904

Supelco

Oxazepam-d5 solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15D5H6ClN2O2
CAS Number:
Molecular Weight:
291.74
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

shipped in

wet ice

storage temp.

−20°C

SMILES string

[2H]c1c([2H])c([2H])c(c([2H])c1[2H])C2=NC(O)C(=O)Nc3ccc(Cl)cc23

InChI

1S/C15H11ClN2O2/c16-10-6-7-12-11(8-10)13(9-4-2-1-3-5-9)18-15(20)14(19)17-12/h1-8,15,20H,(H,17,19)/i1D,2D,3D,4D,5D

InChI key

ADIMAYPTOBDMTL-RALIUCGRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Oxazepam is a benzodiazepine drug used to treat anxiety and insomnia and to control the symptoms of alcohol withdrawal. This stable-labeled internal standard is suitable for isotope dilution methods or the quantitation of oxazepam levels using GC/MS or LC/MS in testing applications such as clinical toxicology, urine drug testing, forensic analysis or pain prescription monitoring.

Application


  • Oxazepam-d5 as an internal standard in toxicology: Oxazepam-d5 is utilized in gas chromatography-tandem mass spectrometry for the determination of lorazepam and its metabolites in plasma and urine, demonstrating its utility as a stable isotope-labeled compound for enhancing analytical specificity and accuracy (Pichini et al., 1999).

  • Stable isotope-labeled Oxazepam for quantification in forensic science: This compound serves as a critical internal standard in sensitive GC-MS procedures for analyzing benzodiazepines and their metabolites in biological specimens, vital for forensic applications (elSohly et al., 1997).

  • Deuterated Oxazepam reference material for mass spectrometry: Oxazepam-d5 is used in the development of analytical methods for the detection of benzodiazepine metabolites, providing high accuracy and precision essential for clinical and toxicological evaluations (Meatherall, 1994).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service