Skip to Content
Merck
All Photos(1)

Key Documents

N-106

Supelco

Naltrexone-3-β-D-glucuronide solution

100 μg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H31NO10
CAS Number:
Molecular Weight:
517.53
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

shipped in

wet ice

storage temp.

−20°C

SMILES string

N2([C@H]3[C@]4([C@@]5([C@@H](Oc6c5c(ccc6O[C@@H]7O[C@@H]([C@H]([C@@H]([C@H]7O)O)O)C(=O)O)C3)C(=O)CC4)CC2)O)CC1CC1

InChI

1S/C26H31NO10/c28-13-5-6-26(34)15-9-12-3-4-14(35-24-19(31)17(29)18(30)21(37-24)23(32)33)20-16(12)25(26,22(13)36-20)7-8-27(15)10-11-1-2-11/h3-4,11,15,17-19,21-22,24,29-31,34H,1-2,5-10H2,(H,32,33)/t15-,17+,18+,19-,21+,22+,24-,25+,26-/m1/s1

InChI key

KCSKQJYZSCIQRR-ODTDCTFRSA-N

General description

A certified solution standard of Naltrexone-3-ß-D-glucuronide, a metabolite of the opiate Naltrexeone. Sold under the trade names Revia, Depade and Vivitrol, naltrexone is prescribed primarily for the management of alcohol or opiate dependence. This analytical reference standard is applicable for use in LC/MS or GC/MS applications such as pharmaceutical research, forensic analysis, clinical toxicology, pain prescription monitoring or urine drug testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Susanne E Pors et al.
Veterinary microbiology, 150(3-4), 354-361 (2011-03-29)
Pasteurella multocida is a widespread respiratory pathogen in pigs associated with atrophic rhinitis and contributing to aggravation of the pulmonary lesions. The aims of the present study were to characterize isolates of P. multocida from porcine bronchopneumonia by pulsed-field gel
Kenneth Viar et al.
PloS one, 15(6), e0235110-e0235110 (2020-06-26)
Programmed axonal degeneration, also known as Wallerian degeneration, occurs in immune-mediated central nervous system (CNS) inflammatory disorders such as multiple sclerosis and the animal model experimental allergic encephalomyelitis (EAE). Sterile alpha and TIR domain containing protein 1 (SARM1) functions to
Najate Benamer et al.
Nature communications, 11(1), 5151-5151 (2020-10-15)
Myelination of projection neurons by oligodendrocytes is key to optimize action potential conduction over long distances. However, a large fraction of myelin enwraps the axons of parvalbumin-positive fast-spiking interneurons (FSI), exclusively involved in local cortical circuits. Whether FSI myelination contributes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service