Skip to Content
Merck
All Photos(1)

Key Documents

N-069

Supelco

(±)-Norepinephrine-D6 hydrochloride solution

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H5D6NO3·HCl
CAS Number:
Molecular Weight:
211.68
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing
clinical testing

format

single component solution

storage temp.

−70°C

SMILES string

Cl.NC(C(O)(c1c(c(c(c(c1[2H])[2H])O)O)[2H])[2H])([2H])[2H]

InChI

1S/C8H11NO3.ClH/c9-4-8(12)5-1-2-6(10)7(11)3-5;/h1-3,8,10-12H,4,9H2;1H/i1D,2D,3D,4D2,8D;

InChI key

FQTFHMSZCSUVEU-GVFLBQLQSA-N

General description

Norepinephrine, also known as noradrenaline, is a marker for catecholamine-secreting tumors such as pheochromocytoma, paraganglioma, and neuroblastoma. This internal standard is suitable for LC-MS/MS monitoring of norepinephrine levels in urine or plasma for diagnostic testing or endocrinology.

Application


  • Neurotransmitter analysis: Norepinephrine-D6 HCl was employed as an internal standard in mass spectrometry for accurate neurotransmitter quantification in bioanalytical studies, aiding in precise measurements critical for neurological research (El-Deeb et al., 2024).

  • Pharmacological research: Research utilized norepinephrine-D6 HCl for tracing metabolic pathways in pharmacology studies, demonstrating its value in discovering new therapeutic targets (Hu et al., 2024).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service