Skip to Content
Merck
All Photos(1)

Documents

M-153

Supelco

Methotrexate-D3 solution

100 μg/mL in methanol with 0.01N NaOH, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H19N8O5D3
CAS Number:
Molecular Weight:
457.46
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol with 0.01N NaOH

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

−20°C

SMILES string

[2H]C([2H])([2H])N(Cc1cnc2nc(N)nc(N)c2n1)c3ccc(cc3)C(=O)NC(CCC(O)=O)C(O)=O

InChI

1S/C20H22N8O5/c1-28(9-11-8-23-17-15(24-11)16(21)26-20(22)27-17)12-4-2-10(3-5-12)18(31)25-13(19(32)33)6-7-14(29)30/h2-5,8,13H,6-7,9H2,1H3,(H,25,31)(H,29,30)(H,32,33)(H4,21,22,23,26,27)/i1D3

InChI key

FBOZXECLQNJBKD-FIBGUPNXSA-N

General description

An internal standard suitable for therapeutic drug monitoring analyses of methotrexate patient serum samples by LC-MS/MS. Used in the treatment of cancer and autoimmune diseases, methotrexate is an antimetabolite and antifolate drug monitored by clinical labs to ensure patients remain within the drug′s therapeutic range. The Snap-N-Spike® solution format allows laboratories to eliminate weighing operations of hazardous substances such as methotrexate, significantly reducing occupational exposure hazards related to handling of acutely toxic powders, and provides the ability to spike into their matrix of choice just before use.

Application


  • Application in leukemia research: Methotrexate-D₃ is utilized as an internal standard in LC-MS/MS analysis to monitor the bioavailability and metabolic pathways of thioguanosine and methylthioinosine in children with acute lymphoblastic leukemia, supporting its use in pediatric oncology and pharmacokinetics studies (Han et al., 2022).

  • Enhancement of drug monitoring techniques: Employed in high-throughput MALDI-isotope dilution mass spectrometry, Methotrexate-D₃ allows for the ultrafast and selective quantification of methotrexate in human plasma, proving essential for optimizing therapeutic regimens in cancer treatment (Meesters et al., 2011).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - Met. Corr. 1 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Matthew Jeronimo et al.
Analytical and bioanalytical chemistry, 407(23), 7083-7092 (2015-07-05)
An effective wipe sampling and LC-MS/MS method was developed to simultaneously analyze six commonly administered antineoplastic drugs in stainless steel surface. The analyzed drugs were methotrexate, paclitaxel, cyclophosphamide, 5-fluorouracil, vincristine, and oxaliplatin, a frequently prepared antineoplastic drug that has not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service