Skip to Content
Merck
All Photos(1)

Key Documents

I-042

Supelco

Infliximab solution

certified reference material, ampule of 0.25 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size

0.25 ML
$558.00

$558.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
0.25 ML
$558.00

About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

$558.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 0.25 mL

manufacturer/tradename

Cerilliant®

technique(s)

liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

General description

Infliximab, a purified chimeric IgG monoclonal antibody protein, is created from recombinant DNA technology. It has both mouse and human components to effectively block the activity of tumor necrosis factor-alpha (TNF-α). Infliximab is used to stimulate the immune system of our body and treat certain diseases, including ulcerative colitis, active rheumatoid arthritis, and ankylosing spondylitis.[1]

This product is a certified reference material, consisting of a pure substance within an aqueous solvent solution, 12.5 mM histidine buffer. This standard is prepared and certified under the ISO 17034, ISO/IEC 17025, and ISO 9001 standards, meeting the requirements of a certified reference material and a primary standard per ISO guidelines. This primary standard is suitable to prepare calibrators and/or controls.

Application

Infliximab (Remicade) solution is suitable for analytical and R&D applications for in vitro identification, calibration, and quantification.

Features and Benefits

  • Infliximab (Remicade) content is measured via quantitative amino acid analysis (AAA).
  • Ready to use solution.
  • Shelf life and retest date is determined by long term stability studies.
  • The standard is created with precise gravimetric preparation techniques, using fully qualified and calibrated balances that adhere to ISO 17025 standards.
  • The density and material certification are traceable to the SI and higher-order reference materials through mass measurement, calibration, and instrument qualification.
  • The AAA, high-performance liquid chromatography (HPLC) ultraviolet (UV)/mass spectrometry (MS) calibration certified techniques establish traceability.
  • Traceability is ensured to higher order standards from NIST through an unbroken chain of comparisons.

Preparation Note

  • Thaw contents at either room or refrigerated temperature and mix well prior to usage.
  • Do not refreeze the product after thawing it.
  • Established good laboratory practices should be used when quantitatively transferring the appropriate volume for spiking into a matrix or for dilution purposes.

Other Notes

This product′s purpose is for testing. THIS PRODUCT IS NOT INTENDED FOR HUMAN OR THERAPEUTIC USE. Please refer to the safety data sheet for specific information regarding any hazards and appropriate precautions to be taken.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Infliximab
StatPearls [Internet] (2023)

Articles

Learn about the separation of several charge variants of two monoclonal antibodies, NISTmAb and Infliximab, using a polymeric cation exchange column and MS-compatible mobile phases.

Characterize mAb monomers, aggregates, and fragments using SEC-UV workflow with Zenix® and Zenix®-C SEC columns.

Questions

  1. Could you kindly furnish additional details regarding your product, specifically regarding its concentration?

    1 answer
    1. The concentration of Infliximab (Remicade) in Product No. I-042 is listed in the lot-specific Certificate of Analysis. A sample Certificate of Analysis can be found in the Documentation section of the product data page. To obtain the Certificate of Analysis for lots currently in stock, Please contact technical service.

      https://www.sigmaaldrich.com/product/cerillian/i042#product-documentation

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service