Skip to Content
Merck
All Photos(1)

Key Documents

I-024

Supelco

Interference Mix 2 (For Qualitative Use Only)

100 μg/μL each analyte in methanol, analytical standard, Cerilliant®

Synonym(s):

Qualitative Interference Mixture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

form

liquid

composition

Gabapentin, 100 μg/mL
Pregabalin, 100 μg/mL
Salicylic acid, 100 μg/mL
Valproic acid, 100 μg/mL
Vigabatrin, 100 μg/mL

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

multi-component solution

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

A multi-component solution standard of routinely monitored over-the-counter (OTC) and prescription drugs gabapentin, pregabalin, salicylic acid, valproic acid and vigabatrin. This standard is for qualitative use only in the evaluation of interferences between the mix components and target analytes of interest in a testing laboratory′s analytical methods.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service