Skip to Content
Merck
All Photos(1)

Documents

G-016

Supelco

Green Tea Catechin Mix

100 μg/mL each component in acetonitrile: water (8:2) with 5% 1M HCl, ampule of 1.0 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10N4O2
Molecular Weight:
194.19
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

feature

SNAP-N-SPIKE®/SNAP-N-SHOOT®

composition

Caffeine
(+)-Catechin
(−)-Catechin 3-gallate
(−)-Epicatechin
(−)-Epicatechin-3-gallate
(−)-Epigallocatechin 3-gallate
(−)-Gallocatechin
(−)-Gallocatechin 3-gallate

packaging

ampule of 1.0 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL each component in acetonitrile: water (8:2) with 5% 1M HCl

application(s)

food and beverages

format

multi-component solution

storage temp.

−20°C

General description

A certified multi-component mix of Caffeine and the common Green Tea catechins: (-)-Epigallocatechin 3-gallate, (+)-Catechin, (-)-Epicatechin, (-)-Epicatechin 3-gallate, (-)-Gallocatechin, (-)-Gallocatechin 3-gallate, and (-)-Catechin 3-gallate. This Snap-N-Shoot® solution mix is suitable for quality control testing of Green Tea beverages or supplements by LC or LC/MS. The Snap-N-Shoot® format and multi-component solution mix offer several advantages from added convenience with less time weighing plant extracts or powders and preparing solutions to improved efficiency from less time rerunning samples or remaking reference solutions due to different raw material purity factors. The sealed ampoule provides additional precautions for storage of these sensitive phytochemicals.


Discover the entire range of phytochemical certified reference material solutions from Cerilliant

Application


  • Uncovering the role of yttrium in a cerium-based binary oxide in the catalytic conversion of carbon dioxide and methanol to dimethyl carbonate.: This study highlights the role of yttrium doping in enhancing the catalytic properties of cerium(IV) oxide for converting carbon dioxide and methanol into dimethyl carbonate, presenting a potential pathway for carbon dioxide utilization (Wang et al., 2023).

  • Studies of adsorption of α,β-unsaturated carbonyl compounds on heterogeneous Au/CeO(2), Au/TiO(2) and Au/SiO(2) catalysts during reduction by hydrogen.: This research investigates the adsorption characteristics of α,β-unsaturated carbonyl compounds on cerium(IV) oxide and other metal oxides, providing insights into the surface interactions during hydrogen reduction processes (Zielinski et al., 2022).

  • Photocatalytic reduction of CO(2) based on a CeO(2) photocatalyst loaded with imidazole fabricated N-doped graphene and Cu(ii) as cocatalysts.: Explores the use of cerium(IV) oxide in a complex photocatalyst system for the reduction of carbon dioxide, aiming to provide a sustainable method for CO2 reduction and energy production (Zhou and Liu, 2017).



Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service