Skip to Content
Merck
All Photos(1)

Key Documents

F-096

Supelco

Flualprazolam-D4 solution

100 μg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$223.00

$223.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$223.00

About This Item

Empirical Formula (Hill Notation):
C17H8D4ClFN4
Molecular Weight:
330.78
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$223.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

format

single component solution

storage temp.

−20°C

SMILES string

FC1=C([2H])C([2H])=C([2H])C([2H])=C1C2=NCC3=NN=C(C)N3C4=C2C=C(Cl)C=C4

InChI key

MPZVLJCMGPYWQQ-QFFDRWTDSA-N

General description

This stable-labeled internal standard is suitable for quantification of flualprazolam by LC/MS or GC/MS for clinical toxicology, forensic analysis, or urine drug testing applications. Flualprazolam, a 2′-fluoro derivative of alprazolam, is a benzodiazepine sold as designer drug on the illicit drug market.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service