Skip to Content
Merck
All Photos(1)

Documents

F-052

Supelco

Furosemide Impurity Mixture solution

5 μg/mL (50:50 Acetonitrile:Water, each analyte), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11ClN2O5S
Molecular Weight:
330.74
UNSPSC Code:
12352200
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

composition

2-Chloro-4-furfurylamino-5-sulfamoylbenzoic acid, 5 μg/mL
4-Chloro-5-sulfamoylanthranilic acid (Furosemide RCB), 5 μg/mL
Furosemide, 5 μg/mL

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

5 μg/mL (50:50 Acetonitrile:Water, each analyte)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

multi-component solution

storage temp.

−20°C

General description

Furosemide is a powerful diuretic agent with the ascending limb of Henle′s loop of nephron as its primary site of action. It works by preventing the reabsorption of sodium and increasing the secretion of renal vasodilatory prostaglandin E. In combination with nitrates, it is used as the standard treatment for pulmonary edema caused by acute heart failure.

Application

Furosemide Impurity Mixture solution may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Furosemide, Pharmaceutical Secondary Standard; Certified Reference Material
    Furosemide Related Compound B, Pharmaceutical Secondary Standard; Certified Reference Material
    Furosemide Related Compound A, Pharmaceutical Secondary Standard; Certified Reference Material

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

50.0 °F

Flash Point(C)

10 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Furosemide effect on isolated perfused tubules
Burg M, et al.
The American Journal of Physiology, 225(1), 119-124 (1973)
Lack of renoprotective effects of dopamine and furosemide during cardiac surgery
Lassnigg A, et al.
Journal of the American Society of Nephrology, 11(1), 97-104 (2000)
Randomised trial of high-dose isosorbide dinitrate plus low-dose furosemide versus high-dose furosemide plus low-dose isosorbide dinitrate in severe pulmonary oedema
Cotter G, et al.
Lancet, 351(9100), 389-393 (1998)
Furosemide
USP42/NF37: United States Pharmacopeia and National Formulary
United States Pharmacopeia/National Formulary, 41(3), 2006-2006 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service