Skip to Content
Merck
All Photos(1)

Documents

ERT-053

Supelco

Thiodiglycol solution

1000 μg/mL in methanol, ampule of 1.2 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H10O2S
CAS Number:
Molecular Weight:
122.19
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1.2 mL

manufacturer/tradename

Cerilliant®

concentration

1000 μg/mL in methanol

application(s)

environmental

format

single component solution

storage temp.

−20°C

SMILES string

OCCSCCO

InChI

1S/C4H10O2S/c5-1-3-7-4-2-6/h5-6H,1-4H2

InChI key

YODZTKMDCQEPHD-UHFFFAOYSA-N

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N G Medvedeva et al.
Izvestiia Akademii nauk. Seriia biologicheskaia, (1)(1), 91-99 (2012-05-10)
Destruction of mustard gas hydrolysis products by bacterial cultures isolated from soils and bottom waters at the sites of chemical weapons disposal has been studied. Among the tested microorganisms, the soil bacteria Pseudomonas putida Y-21 and Rhodococcus erythropolis 8D and
Leila El Bassi et al.
Journal of hazardous materials, 167(1-3), 124-127 (2009-01-31)
Microbial degradation of thiodiglycol (bis(2-hydroxyethyl)sulfide, TDG) with petroleum-desulfurizing soil bacteria was examined. Among the bacteria tested, several strains belonging to the genera Rhodococcus and Gordonia grew on TDG as the sole sulfur source. The selected strain Rhodococcus sp. strain T09
Hong Li et al.
Biodegradation, 24(1), 125-135 (2012-07-04)
Thiodiglycol (TDG) is both the precursor for chemical synthesis of mustard gas and the product of mustard gas hydrolysis. TDG can also react with intermediates of mustard gas degradation to form more toxic and/or persistent aggregates, or reverse the pathway
G Karvaly et al.
Journal of applied toxicology : JAT, 28(1), 21-26 (2007-04-13)
Subcutaneous microdialysis was employed for monitoring thiodiglycol (2,2'-thiodiethanol, TDG) levels with the aim of characterizing the transdermal penetration of topically applied liquid sulfur mustard (2,2'-dichlorodiethyl sulfide, SM) in rats. TDG levels, evaluated in 20 min dialysates collected over a 6
Karolin K Kroening et al.
Analytical and bioanalytical chemistry, 393(8), 1949-1956 (2009-02-14)
Sulfur mustard (HD), bis(2-chloroethyl)sulfide, is one of a class of mustard agents which are chemical warfare agents. The main chemical warfare hydrolysis degradation products of sulfur mustards are: thiodiglycol, bis(2-hydroxyethylthio)methane, 1,2-bis(2-hydroxyethylthio)ethane, 1,3-bis(2-hydroxyethylthio)propane, and 1,4-bis(2-hydroxyethylthio)butane. The aim of this study is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service