Skip to Content
Merck
All Photos(1)

Key Documents

ERH-004S

Supelco

HMX solution

1000 μg/mL in acetonitrile, ampule of 1.2 mL, certified reference material, Cerilliant®

Synonym(s):

1,3,5,7-Tetranitro-1,3,5,7-tetraazacyclooctane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8N8O8
CAS Number:
Molecular Weight:
296.16
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1.2 mL

manufacturer/tradename

Cerilliant®

concentration

1000 μg/mL in acetonitrile

application(s)

environmental

format

single component solution

storage temp.

−20°C

SMILES string

O=[N+]([O-])N1CN([N+]([O-])=O)CN([N+]([O-])=O)CN([N+]([O-])=O)C1

InChI

1S/C4H8N8O8/c13-9(14)5-1-6(10(15)16)3-8(12(19)20)4-7(2-5)11(17)18/h1-4H2

InChI key

UZGLIIJVICEWHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hardiljeet K Boparai et al.
Chemosphere, 79(8), 865-872 (2010-03-17)
Zerovalent iron barriers have become a viable treatment for field-scale cleanup of various ground water contaminants. While contact with the iron surface is important for contaminant destruction, the interstitial pore water within and near the iron barrier will be laden
Guilherme R Lotufo
Ecotoxicology and environmental safety, 74(3), 301-306 (2011-01-25)
Sheepshead minnows (Cyprinodon variegatus) were exposed to radiolabeled isotopes of the explosives 2,4,6-trinitrotoluene (TNT), exahydro-1,3,5-trinitro-1,3,5-triazine (commonly known as RDX), and octahydro-1,3,5,7-tetranitro-1,3,5,7-tetrazocine (commonly known as HMX), yielding the bioconcentration factors (BCF) of 3.3, 0.7, and 0.1 L kg(-1), respectively. For TNT
Chun-jiang An et al.
Journal of hazardous materials, 179(1-3), 526-532 (2010-04-03)
The performance of mesophilic anaerobic granules to degrade octahydro-1,3,5,7-tetranitro-1,3,5,7-tetrazocine (HMX) was investigated under various conditions. The results of batch experiments showed that anaerobic granules were capable of removing HMX from aqueous solution with high efficiency. Both biotic and abiotic mechanisms
Sudeep Perumbakkam et al.
Research in microbiology, 163(8), 567-575 (2012-08-21)
Explosives such as octahydro-1,3,5,7-tetranitro-1,3,5,7-tetrazocine (HMX) provide a challenge in terms of bioremediation. In the present study, sheep rumen was studied for its potential to detoxify HMX using analytical chemistry and molecular microbial ecology tools. Results indicated significant loss (p <
Thomas A Douglas et al.
Environmental toxicology and chemistry, 30(2), 345-353 (2010-11-03)
Potentially toxic nitroaromatic and nitramine compounds are introduced onto soils during detonation of explosives. The present study was conducted to investigate the desorption and transformation of explosive compounds loaded onto three soils through controlled detonation. The soils were proximally detonated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service