Skip to Content
Merck
All Photos(1)

Key Documents

E-080

Supelco

N-Ethylcathinone hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15NO · HCl
CAS Number:
Molecular Weight:
213.70
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland)

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CC(NCC)C(C1=CC=CC=C1)=O.Cl

InChI

1S/C11H15NO.ClH/c1-3-12-9(2)11(13)10-7-5-4-6-8-10;/h4-9,12H,3H2,1-2H3;1H

InChI key

XCVDYVFUJZVVKL-UHFFFAOYSA-N

General description

N-Ethylcathinone, or ethylpropion, is an amphetamine and stimulant drug sold along with mephedrone as “ecstasy” at parties and raves. With street names such as ETH-CAT, the illicit drug is also sold in bath salt blends across the US, Europe, and Australia. This Certified Spiking Solution® is suitable as starting material in calibrators and controls for GC/MS or LC/MS bath salt testing applications in forensic analysis, urine drug testing or clinical toxicology.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service