Skip to Content
Merck
All Photos(1)

Key Documents

D-052

Supelco

N-Desmethyltapentadol solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H21NO
CAS Number:
Molecular Weight:
207.31
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CC[C@H]([C@@H](C)CNC)c1cccc(O)c1

InChI

1S/C13H21NO/c1-4-13(10(2)9-14-3)11-6-5-7-12(15)8-11/h5-8,10,13-15H,4,9H2,1-3H3/t10-,13+/m0/s1

InChI key

PQQINTFVECNXLC-GXFFZTMASA-N

General description

N-Desmethyltapentadol is a primary urinary and blood metabolite of the analgesic tapentadol (Nucynta®). This certified reference solution is suitable for numerous GC/MS or LC/MS applications from pharmaceutical testing and prescription pain monitoring to forensic analysis or clinical toxicology. Tapentadol is prescribed for treatment of both acute and chronic pain.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Nucynta is a registered trademark of Johnson & Johnson
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service