Skip to Content
Merck

Skip To

D-034

Dextrorphan tartrate solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C21H29NO7
CAS Number:
Molecular Weight:
407.46
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
EC Number:
200-659-6
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

description

Drug Control: Kábítószer / Narcotic Drug (Hungary), 78/2022. (XII. 28.) BM rendelet

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Kábítószer / Narcotic Drug (Hungary), 78/2022. (XII. 28.) BM rendelet

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

O[C@H]([C@@H](O)C(O)=O)C(O)=O.CN1CC[C@@]23CCCC[C@@H]2[C@@H]1Cc4ccc(O)cc34

InChI

1S/C17H23NO.C4H6O6/c1-18-9-8-17-7-3-2-4-14(17)16(18)10-12-5-6-13(19)11-15(12)17;5-1(3(7)8)2(6)4(9)10/h5-6,11,14,16,19H,2-4,7-10H2,1H3;1-2,5-6H,(H,7,8)(H,9,10)/t14-,16+,17+;1-,2-/m11/s1

InChI key

RWTWIZDKEIWLKQ-XCPWPWHNSA-N

General description

A Snap-N-Spike® Reference Solution suitable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, P450 trials, or pharmaceutical research. Dextrorphan is a primary metabolite of dextromethorphan in blood. Dextromethorphan, an active ingredient in many over-the-counter cold and cough medicines, is used recreationally as a hallucinogen with effects similar to ketamine and phencyclidine (PCP).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Supply conditions do not apply to the regions and states of Brazil: North, Northeast, Mato Grosso do Sul, Mato Grosso, and Rio Grande do Sul.


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters