Skip to Content
Merck
All Photos(1)

Key Documents

C-244

Supelco

Cannabichromenic Acid (CBCA) solution

1 mg/mL (1% DIPEA and 0.05% Ascorbic acid in Acetonitrile), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$297.00

$297.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$297.00

About This Item

Empirical Formula (Hill Notation):
C22H30O4
CAS Number:
Molecular Weight:
358.47
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$297.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL (1% DIPEA and 0.05% Ascorbic acid in Acetonitrile)

format

single component solution

storage temp.

−70°C

SMILES string

CCCCCC1=C(C(O)=O)C(O)=C2C(OC(CCC=C(C)C)(C)C=C2)=C1

InChI

1S/C22H30O4/c1-5-6-7-10-16-14-18-17(20(23)19(16)21(24)25)11-13-22(4,26-18)12-8-9-15(2)3/h9,11,13-14,23H,5-8,10,12H2,1-4H3,(H,24,25)

InChI key

HRHJHXJQMNWQTF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cannabichromenic acid (CBCA) is a precursor of CBC biosynthesis, a non-psychoactive cannabinoid of Cannabis that reportedly exerts anti-inflammatory, antimicrobial and analgesic activity. This Certified Spiking Solution® is suitable for cannabichromenic acid testing methods by GC/MS, LC/MS or HPLC for applications in testing of Cannabis potency or impurity profiling, pharmaceutical research, and forensic analysis. Cerilliant solution Certified Reference Materials (CRMs) of cannabinoids are supplied in a convenient, quantitative, US DEA-exempt solution format and with TK#s for Canadian customers.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service