Skip to Content
Merck
All Photos(1)

Key Documents

C-181

Supelco

7-Carboxy cannabidiol (7-COOH-CBD) solution

1 mg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H28O4
CAS Number:
Molecular Weight:
344.44
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

OC1=CC(CCCCC)=CC(O)=C1C2C(C(C)=C)CCC(C(O)=O)=C2

InChI

1S/C21H28O4/c1-4-5-6-7-14-10-18(22)20(19(23)11-14)17-12-15(21(24)25)8-9-16(17)13(2)3/h10-12,16-17,22-23H,2,4-9H2,1,3H3,(H,24,25)

Application


  • Pharmacokinetic profiling: 7-Carboxy cannabidiol (7-COOH-CBD) is used in pharmacokinetic studies to understand the metabolic pathways of cannabidiol in human plasma. A validated method facilitates the simultaneous quantification of CBD, THC, and their metabolites, crucial for developing effective cannabinoid therapies (Kevin et al., 2021).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service