Skip to Content
Merck
All Photos(1)

Key Documents

C-129

Supelco

Clobazam-8-chloro isomer-13C6 solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C6C10H13ClN2O2
Molecular Weight:
306.70
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

O=C1CC(N([13C]2=[13CH][13CH]=[13CH][13CH]=[13CH]2)C3=CC=C(Cl)C=C3N1C)=O

InChI

1S/C16H13ClN2O2/c1-18-14-9-11(17)7-8-13(14)19(16(21)10-15(18)20)12-5-3-2-4-6-12/h2-9H,10H2,1H3/i2+1,3+1,4+1,5+1,6+1,12+1

InChI key

CFQSNGPGPDIAQQ-FLBQOMCMSA-N

General description

A stable-labeled internal standard suited for quantification of clobazam levels in urine or other matrices by LC/MS or GC/MS for clinical toxicology, urine drug testing, or forensic analysis applications. Marketed under the brand names Frisium, Urbanol, and Onfi®, clobazam is a benzodiazepine used as an anxiolytic and anticonvulsant.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Onfi is a registered trademark of Lundbeck LLC
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service