Skip to Content
Merck
All Photos(1)

Documents

C-039

Supelco

Codeine-D3 solution

ampule of 5 mL, certified reference material, 100 μg/mL in methanol, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18D3NO3
CAS Number:
Molecular Weight:
302.38
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

certified reference material

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 5 mL

manufacturer/tradename

Cerilliant®

drug control

estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

100 μg/mL in methanol

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

−20°C

SMILES string

[2H]C([2H])([2H])N1CC[C@]23[C@H]4Oc5c(OC)ccc(C[C@@H]1[C@@H]2C=C[C@@H]4O)c35

InChI

1S/C18H21NO3/c1-19-8-7-18-11-4-5-13(20)17(18)22-16-14(21-2)6-3-10(15(16)18)9-12(11)19/h3-6,11-13,17,20H,7-9H2,1-2H3/t11-,12+,13-,17-,18-/m0/s1/i1D3

InChI key

OROGSEYTTFOCAN-SBGSAQJDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Codeine-D3 is a stable-labeled internal standard suitable for many LC/MS or GC/MS testing applications including pain prescription monitoring, forensic analysis, clinical toxicology, urine drug testing or isotope dilution methods. Codeine is an opiate used for its analgesic, antitussive, and sedative properties and is the second-most predominant alkaloid in opium. Since opiate drugs are classified as controlled substances, Cerilliant offers most opiate reference standards as DEA-exempt solutions for added convenience.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service