Skip to Content
Merck
All Photos(1)

Key Documents

B-023

Supelco

BSTFA

with 1% TMCS, kit of 10 × 1 mL, analytical standard, Cerilliant®

Synonym(s):

N,O-Bis(trimethylsilyl)trifluoroacetamide, BSTFA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C[=NSi(CH3)3]OSi(CH3)3
CAS Number:
Molecular Weight:
257.40
Beilstein:
2050024
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

Product Name

BSTFA (with 1% TMCS), kit of 10 × 1 mL, analytical standard, Cerilliant®

grade

analytical standard

Quality Level

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

packaging

kit of 10 × 1 mL

manufacturer/tradename

Cerilliant®

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.384 (lit.)

bp

45-50 °C/14 mmHg (lit.)

density

0.969 g/mL at 25 °C (lit.)

application(s)

forensics and toxicology

format

neat

storage temp.

−20°C

SMILES string

C[Si](C)(C)O\C(=N\[Si](C)(C)C)C(F)(F)F

InChI

1S/C8H18F3NOSi2/c1-14(2,3)12-7(8(9,10)11)13-15(4,5)6/h1-6H3/b12-7+

InChI key

XCOBLONWWXQEBS-KPKJPENVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

BSTFA (with 1% TMCS) may be used as a derivatizing agent in the following:
  • Analysis of estrogenic compounds in water samples using gas chromatography coupled to mass spectrometry (GC-MS).[1]
  • Quantification of α-pyrrolidinovalerophenone (α-PVP) and its metabolites in urine samples using gas chromatography coupled to mass spectrometry (GC-MS).[2]

Features and Benefits

  • BSTFA is very versatile, reacting with a range of polar organic compounds and replacing active hydrogens with a −Si(CH3)3 (trimethylsilyl) group.
  • Reacts rapidly and more completely than BSA.;
  • TMCS increases the reactivity of BSTFA (or other silylation reagents).
  • Amides and many secondary amines and hindered hydroxyls, incompletely derivatized by BSTFA alone, can be derivatized by adding 1-20% TMCS to BSTFA.
  • Has good solvent properties and can function as a silylation reagent without additional solvents.
  • Alternatively, the mixture is very soluble in most commonly used silylation solvents.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

75.2 °F - closed cup

Flash Point(C)

24.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optimisation of derivatisation for the analysis of estrogenic compounds in water by solid-phase extraction gas chromatography--mass spectrometry
Zhang LZ, et al.
Analytica Chimica Acta, 577(1), 52-61 (2006)
Determination of synthetic cathinone $\alpha$-pyrrolidinovalero-phenone and its metabolite in urine using solid-phase extraction and gas chromatography--mass spectrometry
Cheng WK, et al.
Rapid Communications in Mass Spectrometry, 34(1), e8579-e8579 (2020)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service