Skip to Content
Merck
All Photos(1)

Documents

A-130

Supelco

Acetyl fentanyl-13C6 solution

50 μg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®

Synonym(s):

Deuterium-Labeled Acetyl Fentanyl Solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C6C15H26N2O
Molecular Weight:
328.40
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

50 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

InChI

1S/C21H26N2O/c1-18(24)23(20-10-6-3-7-11-20)21-13-16-22(17-14-21)15-12-19-8-4-2-5-9-19/h2-11,21H,12-17H2,1H3

InChI key

FYIUUQUPOKIKNI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetyl fentanyl is a designer drug and fentanyl analog with a potency 40 times greater than heroin and 80 times greater than morphine. Acetyl fentanyl has been directly implicated in numerous opiate overdose deaths throughout the United States. This stable-labeled internal standard is suitable for quantification of acetyl fentanyl levels in urine or other matrices by LC/MS or GC/MS for clinical toxicology, forensic analysis, prescription monitoring, or urine drug testing applications. This Certified Spiking Solution® is supplied in a convenient, quantitative, US DEA-exempt solution format and with TK #s for Canadian customers.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marwen Moussa et al.
Applied and environmental microbiology, 73(20), 6508-6518 (2007-09-04)
The relationship between membrane permeability, changes in ultrastructure, and inactivation in Escherichia coli strain K-12TG1 cells subjected to high hydrostatic pressure treatment at room and subzero temperatures was studied. Propidium iodide staining performed before and after pressure treatment made it
Pilar Mañas et al.
Applied and environmental microbiology, 70(3), 1545-1554 (2004-03-10)
The relationship between a loss of viability and several morphological and physiological changes was examined with Escherichia coli strain J1 subjected to high-pressure treatment. The pressure resistance of stationary-phase cells was much higher than that of exponential-phase cells, but in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service