A-127
Multicomponent Alcohol Mix
in H2O (varied conc.), certified reference material, Cerilliant®
Synonym(s):
Multi-component Alcohol Mix
About This Item
Recommended Products
grade
certified reference material
Quality Level
form
liquid
feature
Snap-N-Spike®/Snap-N-Shoot®
packaging
ampule of 6 × 1.2 mL
manufacturer/tradename
Cerilliant®
concentration
in H2O (varied conc.)
technique(s)
gas chromatography (GC): suitable
liquid chromatography (LC): suitable
application(s)
forensics and toxicology
format
multi-component solution
storage temp.
2-8°C
InChI
1S/C15H13N3O/c16-11-6-7-13-12(8-11)15(17-9-14(19)18-13)10-4-2-1-3-5-10/h1-8H,9,16H2,(H,18,19)/i1D,2D,3D,4D,5D
InChI key
OYOUQHVDCKOOAL-RALIUCGRSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
related product
Storage Class Code
10 - Combustible liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service