Skip to Content
Merck
All Photos(1)

Key Documents

A-120

Supelco

Aldosterone-13C3 solution

10 μg/mL in acetonitrile, certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$449.00

$449.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$449.00

About This Item

Empirical Formula (Hill Notation):
C1813C3H28O5
Molecular Weight:
363.42
UNSPSC Code:
41116107
NACRES:
NA.24

$449.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

10 μg/mL in acetonitrile

format

single component solution

storage temp.

−20°C

General description

This stable labeled internal standard is suitable for quantitation of aldosterone levels using LC/MS or GC/MS methods for clinical diagnostic applications. Aldosterone is a hormone in the mineralocorticoid family which stimulates sodium transport across cell membranes and assists in the maintenance of blood pressure and blood volume. Aldosterone levels in serum/plasma and urine are measured by LC-MS/MS for investigation of primary or secondary aldosteronism ranging from adrenal cortical hyperplasia to renovascular disease and Bartter′s syndrome.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service