Skip to Content
Merck
All Photos(1)

Documents

A-099

Supelco

Androsterone-d4 glucuronide solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Androsterone-D4 glucuronide solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25D4H34O8
Molecular Weight:
470.59
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

O=C1CC[C@@]2([H])[C@]3([H])CC[C@@]4([H])C([2H])([2H])[C@H](O[C@@H]5O[C@H](C(O)=O)[C@@H](O)[C@H](O)[C@H]5O)C([2H])([2H])C[C@]4(C)[C@@]3([H])CC[C@@]21C

InChI

1S/C25H38O8/c1-24-9-7-13(32-23-20(29)18(27)19(28)21(33-23)22(30)31)11-12(24)3-4-14-15-5-6-17(26)25(15,2)10-8-16(14)24/h12-16,18-21,23,27-29H,3-11H2,1-2H3,(H,30,31)/t12-,13+,14-,15-,16-,18-,19-,20+,21-,23+,24-,25-/m0/s1/i7D2,11D2

InChI key

VFUIRAVTUVCQTF-RDSFDFKRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

An internal standard suitable for LC/MS or GC/MS applications with androsterone and androsterone glucuronide such as diagnostic testing for diagnosis and treatment of gonadal and adrenal diseases or sports testing to monitor testosterone doping. This Certified Spiking Solution® provides added value in allowing for quantitation of androsterone glucuronide levels by LC-MS/MS without the need for hydrolysis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTIONS is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service