Skip to Content
Merck
All Photos(1)

Key Documents

LM1002

Avanti

17:0-20:4 PC

Avanti Research - A Croda Brand LM1002, methanol solution

Synonym(s):

1-heptadecanoyl-2-(5Z,8Z,11Z,14Z-eicosatetraenoyl)-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$253.00

$253.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$253.00

About This Item

Empirical Formula (Hill Notation):
C45H82NO8P
CAS Number:
Molecular Weight:
796.11
UNSPSC Code:
12352211
NACRES:
NA.25

$253.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

methanol solution

packaging

pkg of 1 × 1 mL (LM1002-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand LM1002

concentration

~10 μg/mL (Refer to C of A for lot specific concentration.)

application(s)

lipidomics
metabolomics

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCC/C=C\C/C=C\C/C=C\C/C=C\CCCCC)=O)COC(CCCCCCCCCCCCCCCC)=O)=O

InChI

1S/C45H82NO8P/c1-6-8-10-12-14-16-18-20-22-23-24-26-28-30-32-34-36-38-45(48)54-43(42-53-55(49,50)52-40-39-46(3,4)5)41-51-44(47)37-35-33-31-29-27-25-21-19-17-15-13-11-9-7-2/h14,16,20,22,24,26,30,32,43H,6-13,15,17-19,21,23,25,27-29,31,33-42H2,1-5H3/b16-14-,2

InChI key

MHUZXLUKTLNHIX-GUNPMBPGSA-N

General description

Phosphatidylcholine (PC) is the most abundant phospholipid in eukaryotes and a vital component of the mucosal layer of the colon.[1] It is a strong bilayer-forming lipid.[1]

Application

17:0-20:4 PC has been used as an internal standard for the quantification of dipalmitoylphosphatidylcholine (DPPC) and palmitoyl-oleoyl-phosphatidylcholine (POPC)[2], and as a quantitative lipid standard to monitor various lipid extracts concentration.[3]

Biochem/physiol Actions

Phosphatidylcholine (PC) plays a vital role as a surfactant within the mucus to form a hydrophobic surface to prevents bacterial penetrance.[4] It prevents the accumulation of triglycerides and the formation of lipolysis-resistant lipid droplets.[5]

Packaging

2 mL Amber Glass Sealed Ampule (LM1002-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Integrative Medicine - E-Book (2017)
The effect of exhalation flow on endogenous particle emission and phospholipid composition
Larsson P, et al.
Respiratory Physiology & Neurobiology, 243, 39-46 (2017)
The Membranes of Cells (2016)
Phosphatidylcholine Synthesis for Lipid Droplet Expansion Is Mediated by Localized Activation of CTP Phosphocholine Cytidylyltransferase
Krahmer N, et al.
Cell Metabolism, 14(4), 504-515 (2011)
A comprehensive method for lipid profiling by liquid chromatography-ion cyclotron resonance mass spectrometry
Fauland A, et al.
Journal of Lipid Research, 52(12), 2314-2322 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service