Skip to Content
Merck
All Photos(1)

Key Documents

900210O

Avanti

Polyprenol (13~21)

Avanti Research - A Croda Brand 900210O

Synonym(s):

Polyprenol mixture (13~21)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$199.00

$199.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$199.00

About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

$199.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

liquid

packaging

pkg of 1 × 1 mg (900210O-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 900210O

lipid type

neutral glycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

CC(C)=CCC/C(C)=C/CC/C(C)=C/CC/C(C)=C\CC/C(C)=C\CO

InChI key

YHTCXUSSQJMLQD-PSTDWBAXSA-N

General description

Polyprenols or poly-isoprene-ols are primary monohydric alcohols containing 5 to 25 isoprene units in its carbon skeleton, with a hydroxyl group placed at the end.[1][2] They are found in limited quantities in several plant tissues.[3] Dolichol is an α-saturated polyisoprenoid alcohol with 13-21 isoprene units.[4] Dolichol found in yeast and fungi, facilitate the synthesis of mannoproteins.[5]

Biochem/physiol Actions

Polyprenols can be predominantly used as a natural bioregulators.[3] Dolichol concentration in animal tissues increases with age. Therefore, it can be used as a potential biomarker of aging. Increased serum dolichol levels leads to the development of chronic cholestatic liver diseases.[6]

Packaging

5 mL Clear Glass Sealed Ampule (900210O-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biochemistry (2016)
Carbohydrates Volume 2 of Methods in Plant Biochemistry (2012)
Chromatography of long chain alcohols (polyprenols) from animal and plant sources
Rezanka T and Votruba J
Journal of Chromatography A, 936(1-2), 95-110 (2001)
Biochemistry of Lipids: Biochemistry Series One, 4 (2014)
Accumulation of dolichol in older tissues satisfies the proposed criteria to be qualified a biomarker of aging
Parentini I, et al.
The Journals of Gerontology. Series A, Biological Sciences and Medical Sciences, 60(1), 39-43 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service