Skip to Content
Merck
All Photos(1)

Key Documents

890895P

Avanti

18:1 TAP (DOTAP, MS Salt)

Avanti Research - A Croda Brand 890895P, powder

Synonym(s):

1,2-dioleoyl-3-trimethylammonium-propane (methyl sulfate salt)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$284.00
200 MG
$591.00

$284.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$284.00
200 MG
$591.00

About This Item

Empirical Formula (Hill Notation):
C43H83NO8S
CAS Number:
Molecular Weight:
774.19
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$284.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 2 × 100 mg (890895P-200mg)
pkg of 1 × 25 mg (890895P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 890895P

lipid type

cationic lipids
transfection

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H]C(C[N+](C)(C)C)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.O=S([O-])(OC)=O

InChI

1S/C42H80NO4.CH4O4S/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-41(44)46-39-40(38-43(3,4)5)47-42(45)37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2;1-5-6(2,3)4/h20-23,40H,6-19,24-39H2,1-5H3;1H3,(H,2,3,4)/q+1;/p-1/b22-20-,23-21-;

InChI key

RSMRWWHFJMENJH-LQDDAWAPSA-M

Biochem/physiol Actions

1,2-dioleoyl-3-trimethylammonium-propane (DOTAP) is a cationic lipid, used for in vitro and in vivo nucleic acid and protein delivery.[1] The lipid composition and cell type affects the transfection potential of DOTAP.[2] DOTAP mediates gene knockdown technique in hematopoietic stem cells (HSCs).[3]

Packaging

5 mL Clear Glass Sealed Ampule (890895P-200mg)
5 mL Clear Glass Sealed Ampule (890895P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sabata Martino et al.
Journal of biomedicine & biotechnology, 2009, 410260-410260 (2009-06-09)
RNA interference technology is an ideal strategy to elucidate the mechanisms associated with human CD34(+) hematopoietic stem cell differentiation into dendritic cells. Simple manipulations in vitro can unequivocally yield alloreactive or tolerogenic populations, suggesting key implications of biochemical players that
Dmitri Simberg et al.
Critical reviews in therapeutic drug carrier systems, 21(4), 257-317 (2005-01-11)
Cationic lipid-mediated nucleic acid and protein delivery is becoming increasingly popular for in vitro and in vivo applications. While the chemistry of cationic lipid carriers is moving very rapidly, and more and more sophisticated molecules are being developed, it is
Bieong-Kil Kim et al.
Biochimica et biophysica acta, 1848(10 Pt A), 1996-2001 (2015-06-27)
The effects of lipid compositions on their physicochemical properties and transfection efficiencies were investigated. Four liposome formulations with different 1,2-dioleoyl-3-trimethylammoniumpropane (DOTAP) to dioleoylphosphatidylethanolamine (DOPE) weight ratios were investigated, that is, weight ratios 1:0 (T1P0), 3:1 (T3P1), 1:1 (T1P1), and 1:3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service