Skip to Content
Merck
All Photos(1)

Key Documents

880420P

Avanti

18:0 PEG350 PE

Avanti Research - A Croda Brand 880420P, powder

Synonym(s):

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-350] (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$408.00
500 MG
$2,000.00

$408.00


Estimated to ship onMarch 29, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$408.00
500 MG
$2,000.00

About This Item

CAS Number:
UNSPSC Code:
51191904
NACRES:
NA.25

$408.00


Estimated to ship onMarch 29, 2025


Request a Bulk Order

form

powder

packaging

pkg of 1 × 25 mg (880420P-25mg)
pkg of 1 × 500 mg (880420P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880420P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(OCCOCCOCCOCCOCCOCCOCCOC)=O)=O)(OC(CCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O.[NH4+]

Application

18:0 PEG350 PE is suitable for liposome modification.[1]

Packaging

30 mL Amber Wide Mouth Bottle with Screw Cap (880420P-500mg)
5 mL Clear Glass Sealed Ampule (880420P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zebrafish as a predictive screening model to assess macrophage clearance of liposomes in vivo
Sieber S, et al.
Nanomedicine: Nanotechnology, Biology, and Medicine, 17, 82-93 (2019)
Liming Deng et al.
Biomaterials science, 8(3), 858-870 (2019-12-07)
Hypoxic resistance, photosensitizer toxicity, and target deficiency are major challenges strongly inhibiting the efficacy of clinical photodynamic therapy (PDT) in tumor treatment. To overcome these challenges, we synthesized IR780 and catalase co-loaded liposomes to form a tumor-targeted bio-nanoreactor (LIP-IR-CAT). The
Sandro Sieber et al.
Nanomedicine : nanotechnology, biology, and medicine, 17, 82-93 (2019-01-20)
Macrophage recognition of nanoparticles is highly influenced by particle size and surface modification. Due to the lack of appropriate in vivo screening models, it is still challenging and time-consuming to characterize and optimize nanomedicines regarding this undesired clearance mechanism. Therefore

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service