Skip to Content
Merck
All Photos(1)

Key Documents

880234C

Avanti

DOPE-PEG(2000) Amine

Avanti Research - A Croda Brand

Synonym(s):

DOPE-PEG(2000) Amine ammonium salt, 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-[amino(polyethylene glycol)-2000] ammonium salt

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$254.00

$254.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$254.00

About This Item

Empirical Formula (Hill Notation):
C132H262N3O54P
CAS Number:
Molecular Weight:
2786.45
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$254.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

chloroform solution

packaging

package of 1 × 10 mg (pkg of 1 x10mg (880234C-10MG))

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

10 mg/mL in chloroform

application(s)

advanced drug delivery

lipid type

polymerizable lipids

shipped in

dry ice

storage temp.

−20°C

General description

DOPE-PEG(2000) Amine is a synthetic phospholipid-polyethylene glycol (PEG) conjugate. consisting of dioleoylphosphatidylethanolamine (DOPE), linked to a PEG molecule with a molecular weight of approximately 2000 Daltons, terminated with an amine group. It is commonly used for in both targeted drug delivery and liposomal drug delivery systems research.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service