Skip to Content
Merck
All Photos(1)

Key Documents

872120C

Avanti

18:1 PE-N-16:0

Avanti Research - A Croda Brand

Synonym(s):

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-palmitoyl (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$106.00
10 MG
$179.00

$106.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$106.00
10 MG
$179.00

About This Item

Empirical Formula (Hill Notation):
C57H111N2O9P
CAS Number:
Molecular Weight:
999.47
UNSPSC Code:
12352211
NACRES:
NA.25

$106.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99% (TLC)

form

chloroform solution

packaging

pkg of 1 × 10 mg (872120C-10mg)
pkg of 1 × 5 mg (872120C-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

10 mg/mL (872120C-10MG)
5 mg/mL (872120C-5MG)

lipid type

bioactive lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCCNC(CCCCCCCCCCCCCCC)=O)(OC[C@]([H])(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O)=O.[NH4+]

InChI

1S/C57H108NO9P.H3N/c1-4-7-10-13-16-19-22-25-27-30-33-36-39-42-45-48-56(60)64-52-54(67-57(61)49-46-43-40-37-34-31-28-26-23-20-17-14-11-8-5-2)53-66-68(62,63)65-51-50-58-55(59)47-44-41-38-35-32-29-24-21-18-15-12-9-6-3;/h25-28,54H,4-24,29-53H2,1-3H3,(H,58,59)

InChI key

HRSBIWJTZUDIDL-ZNNVHAOSSA-N

Packaging

5ML Clear Glass Sealed Ampule (872120C-10MG)
5ML Clear Glass Sealed Ampule (872120C-5MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service