Skip to Content
Merck
All Photos(1)

Key Documents

870720P

Avanti

20:0 Coenzyme A

Avanti Research - A Croda Brand 870720P, powder

Synonym(s):

arachidoyl Coenzyme A (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$567.00

$567.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$567.00

About This Item

Empirical Formula (Hill Notation):
C41H83N10O17P3S
CAS Number:
Molecular Weight:
1113.14
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$567.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 5 mg (870720P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870720P

application(s)

lipidomics

lipid type

coenzymes

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@](C(NCCC(NCCSC(CCCCCCCCCCCCCCCCCCC)=O)=O)=O)(C(C)(COP([O-])(OP([O-])(OC[C@H]([C@H]1OP([O-])(O)=O)O[C@H]([C@@H]1O)N2C3=C(C(N)=NC=N3)N=C2)=O)=O)C)[H].[NH4+].[NH4+].[NH4+]

InChI

1S/C41H74N7O17P3S.3H3N/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-32(50)69-25-24-43-31(49)22-23-44-39(53)36(52)41(2,3)27-62-68(59,60)65-67(57,58)61-26-30-35(64-66(54,55)56)34(51)40(63-30)48-29-47-33-37(42)45-28-46-38(33)48;;;/h28-30,34-36,40,51-52H,4-27H2,1-3H3,(H,43,49)(H,44,53)(H,57,58)(H,59,60)(H2,42,45,46)(H2,54,55,56);3*1H3/t30-,34?,35+,36+,40-;;;/m1.../s1

InChI key

FCQFGWHSMZFGCX-LEEULGQNSA-N

General description

20:0 Coenzyme A, also known as arachidoyl Coenzyme A belongs to the group of long-chain acyl-CoAs. It serves as a key lipid metabolite.[1]

Application

20:0 Coenzyme A has been used as an internal standard for measuring long-chain acyl-coenzyme A concentrations and enrichment by liquid chromatography/tandem mass spectrometry.[1] It has also been used as a substrate in ceramide synthase assay.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (870720P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katharina Vollheyde et al.
Lipids, 55(5), 479-494 (2020-05-21)
Wax esters (WE) belong to the class of neutral lipids. They are formed by an esterification of a fatty alcohol and an activated fatty acid. Dependent on the chain length and desaturation degree of the fatty acid and the fatty
Agnieszka U Blachnio-Zabielska et al.
Rapid communications in mass spectrometry : RCM, 25(15), 2223-2230 (2011-07-08)
Long-chain acyl-coenzymes A (acyl-CoAs) (LCACoA) are the activated forms of long-chain fatty acids and serve as key lipid metabolites. Excess accumulation of intracellular LCACoA, diacylglycerols (DAGs) and ceramides may create insulin resistance with respect to glucose metabolism. We present a
Evgeny V Berdyshev et al.
The Journal of biological chemistry, 284(9), 5467-5477 (2009-01-03)
Novel immunomodulatory molecule FTY720 is a synthetic analog of myriocin, but unlike myriocin FTY720 does not inhibit serine palmitoyltransferase. Although many of the effects of FTY720 are ascribed to its phosphorylation and subsequent sphingosine 1-phosphate (S1P)-like action through S1P(1,3-5) receptors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service