Skip to Content
Merck
All Photos(1)

Key Documents

870706P

Avanti

06:0 Coenzyme A

Avanti Research - A Croda Brand 870706P, powder

Synonym(s):

hexanoyl Coenzyme A (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$567.00

$567.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$567.00

About This Item

Empirical Formula (Hill Notation):
C27H55N10O17P3S
CAS Number:
Molecular Weight:
916.77
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$567.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 5 mg (870706P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870706P

application(s)

lipidomics

lipid type

coenzymes

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@](C(NCCC(NCCSC(CCCCC)=O)=O)=O)(C(C)(COP([O-])(OP([O-])(OC[C@H]([C@H]1OP([O-])(O)=O)O[C@H]([C@@H]1O)N2C3=C(C(N)=NC=N3)N=C2)=O)=O)C)[H].[NH4+].[NH4+].[NH4+]

InChI

1S/C27H46N7O17P3S.3H3N/c1-4-5-6-7-18(36)55-11-10-29-17(35)8-9-30-25(39)22(38)27(2,3)13-48-54(45,46)51-53(43,44)47-12-16-21(50-52(40,41)42)20(37)26(49-16)34-15-33-19-23(28)31-14-32-24(19)34;;;/h14-16,20-22,26,37-38H,4-13H2,1-3H3,(H,29,35)(H,30,39)(H,43,44)(H,45,46)(H2,28,31,32)(H2,40,41,42);3*1H3/t16-,20?,21+,22+,26-;;;/m1.../s1

InChI key

MXEFXWIYODKXEJ-WSHYHEJESA-N

Application

06:0 Coenzyme A has been used as an internal standard in ultra-high performance liquid chromatography–tandem mass spectrometry for quantitative analysis of fatty-acyl-Coenzyme A species in biological samples.[1]

Biochem/physiol Actions

06:0 Coenzyme A, also known as hexanoyl coenzyme A, is a derivate of short-chain fatty acid hexanoate.[2] It might be an intermediate of fatty acid metabolism. It acts as a source of N-acyl chain in the synthesis of N-hexanoyl-L-homoserine lactone (HHL) by LuxI homologue RhlI(VsmI).[3] Hexanoyl coenzyme A also acts as a precursor for the synthesis of olivetolic acid (OLA), a polyketide metabolite.[4] N-hexanoyl-CoA is preferred over N-octanoyl-CoA as an acyl donor substrate by GOAT (ghrelin O-acyltransferase).[5]

Packaging

5 mL Amber Glass Screw Cap Vial (870706P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

László Abrankó et al.
Journal of chromatography. A, 1534, 111-122 (2018-01-02)
Fatty acyl-Coenzyme A species (acyl-CoAs) are key biomarkers in studies focusing on cellular energy metabolism. Existing analytical approaches are unable to simultaneously detect the full range of short-, medium-, and long-chain acyl-CoAs, while chromatographic limitations encountered in the analysis of
Futoshi Taura et al.
FEBS letters, 583(12), 2061-2066 (2009-05-21)
Alkylresorcinol moieties of cannabinoids are derived from olivetolic acid (OLA), a polyketide metabolite. However, the polyketide synthase (PKS) responsible for OLA biosynthesis has not been identified. In the present study, a cDNA encoding a novel PKS, olivetol synthase (OLS), was
Y Jiang et al.
Molecular microbiology, 28(1), 193-203 (1998-05-21)
In Pseudomonas aeruginosa, synthesis of the quorum-sensing signal molecules N-butanoyl-L-homoserine lactone (BHL) and N-hexanoyl-L-homoserine lactone (HHL) requires the Luxl homologue Rhll(Vsml). By using thin-layer chromatography in conjunction with high-performance liquid chromatography (HPLC) and mass spectrometry, we show that purified Rhll
Hideko Ohgusu et al.
Biochemical and biophysical research communications, 386(1), 153-158 (2009-06-09)
Ghrelin is a peptide hormone in which serine 3 is modified by n-octanoic acid through GOAT (ghrelin O-acyltransferase). However, the enzymological properties of GOAT remain to be elucidated. We analyzed the in vitro activity of GOAT using the recombinant enzyme.
Jake M Stout et al.
The Plant journal : for cell and molecular biology, 71(3), 353-365 (2012-02-23)
The psychoactive and analgesic cannabinoids (e.g. Δ(9) -tetrahydrocannabinol (THC)) in Cannabis sativa are formed from the short-chain fatty acyl-coenzyme A (CoA) precursor hexanoyl-CoA. Cannabinoids are synthesized in glandular trichomes present mainly on female flowers. We quantified hexanoyl-CoA using LC-MS/MS and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service