Skip to Content
Merck
All Photos(1)

Key Documents

870127P

Avanti

16:0 hexynoyl PE

powder

Synonym(s):

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-(5-hexynoyl) (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$203.00

$203.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$203.00

About This Item

Empirical Formula (Hill Notation):
C43H83N2O9P
CAS Number:
Molecular Weight:
803.10
UNSPSC Code:
12352211
NACRES:
NA.25

$203.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

form

powder

packaging

pkg of 1 × 25 mg (870127P-25mg)

lipid type

headgroup modified lipids
click reagents

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCN([H])C(CCCC#C)=O)=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[NH4+]

InChI

1S/C43H80NO9P.H3N/c1-4-7-10-12-14-16-18-20-22-24-26-28-31-34-42(46)50-38-40(39-52-54(48,49)51-37-36-44-41(45)33-30-9-6-3)53-43(47)35-32-29-27-25-23-21-19-17-15-13-11-8-5-2;/h3,40H,4-5,7-39H2,1-2H3,(H,44,45)(H,48,49);1H3/t40-;/m1./s1

InChI key

YTZUQHQKTCVMFU-YPMTVOEDSA-N

Packaging

5 mL Amber Glass Screw Cap Vial (870127P-25mg)

Storage Class Code

11 - Combustible Solids

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service