Skip to Content
Merck
All Photos(1)

Key Documents

860950P

Avanti

Mito-So

Avanti Research - A Croda Brand (860950P), powder

Synonym(s):

Mito-caged sphingosine

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$506.00
5 MG
$1,510.00

$506.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$506.00
5 MG
$1,510.00

About This Item

Empirical Formula (Hill Notation):
C55H69F3N3O9P
CAS Number:
Molecular Weight:
1004.12
UNSPSC Code:
12352211
NACRES:
NA.25

$506.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Assay

99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (860950P-1mg)
pkg of 1 × 5 mg (860950P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand (860950P)

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

General description

Mito-caged sphingosine (Mito-So) is a mitochondria specific probe. It is a potent agent for determining the sphingosine levels.[1]

Packaging

5ml Clear Glass Sealed Ampule

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mitochondria-specific photoactivation to monitor local sphingosine metabolism and function
Feng S, et al.
eLife, 7, e34555-e34555 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service