Skip to Content
Merck
All Photos(1)

Key Documents

860851P

Avanti

CER9 (t18:0/26:0/18:1)

Avanti Research - A Croda Brand 860851P, powder

Synonym(s):

N-[26-oleoyloxy hexacosanoyl]-D-ribo-phytosphingosine

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$560.00

$560.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$560.00

About This Item

Empirical Formula (Hill Notation):
C62H121NO6
CAS Number:
Molecular Weight:
976.63
UNSPSC Code:
12352211
NACRES:
NA.25

$560.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 mg (860851P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860851P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=C(N[C@H]([C@H](O)[C@H](O)CCCCCCCCCCCCCC)CO)CCCCCCCCCCCCCCCCCCCCCCCCCOC(CCCCCCC/C=C\CCCCCCCC)=O

InChI

1S/C62H121NO6/c1-3-5-7-9-11-13-15-17-27-31-35-39-43-47-51-55-61(67)69-56-52-48-44-40-36-32-29-26-24-22-20-18-19-21-23-25-28-30-34-38-42-46-50-54-60(66)63-58(57-64)62(68)59(65)53-49-45-41-37-33-16-14-12-10-8-6-4-2/h17,27,58-59,62,64-65,68H,3-16,18-26,28-57

InChI key

CEQMSSYIQDMJAU-VUVYNQOZSA-N

Packaging

5 mL Amber Glass Screw Cap Vial (860851P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service