Skip to Content
Merck
All Photos(1)

Key Documents

860819P

Avanti

20:0(2R-OH) Ceramide

Avanti Research - A Croda Brand 860819P, powder

Synonym(s):

N-(2′-(R)-hydroxyarachidoyl)-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$456.00

$456.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$456.00

About This Item

Empirical Formula (Hill Notation):
C38H75NO4
CAS Number:
Molecular Weight:
610.01
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$456.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 5 mg (860819P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860819P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCC/C=C/[C@@H](O)[C@@H](NC([C@H](O)CCCCCCCCCCCCCCCCCC)=O)CO

InChI

1S/C38H75NO4/c1-3-5-7-9-11-13-15-17-18-19-21-23-25-27-29-31-33-37(42)38(43)39-35(34-40)36(41)32-30-28-26-24-22-20-16-14-12-10-8-6-4-2/h30,32,35-37,40-42H,3-29,31,33-34H2,1-2H3,(H,39,43)/b32-30+/t35-,36+,37+/m0/s1

InChI key

CFYLSUZUMMPFNC-PLHXSOCRSA-N

General description

Ceramides containing 2-hydroxy fatty acids (hFA) group are mainly found in the epidermis.[1] 20:0(2R-OH) Ceramide is a 2R hydroxylated arachidic acid-containing ceramide.[2]

Biochem/physiol Actions

Hydroxy fatty acid (hFA)-sphingolipids play a vital role in cell signaling. hFA-ceramides contribute to the permeability barrier function of the surface epithelium of the skin. Fatty acid 2-hydroxylase (FA2H) catalyzes the synthesis of hFA-ceramides. Impaired function of FA2H causes nervous system disorders leukodystrophy and spastic paraparesis in humans.[3] hFA-ceramides are essential for PM02734, an antitumor drug to exhibit its activity.[4]

Packaging

5 mL Amber Glass Screw Cap Vial (860819P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Levels of SCS7/FA2H-mediated fatty acid 2-hydroxylation determine the sensitivity of cells to antitumor PM02734
Herrero AB, et al.
Cancer research, 68(23), 9779-9787 (2008)
Normal fur development and sebum production depends on fatty acid 2-hydroxylase expression in sebaceous glands
Maier H, et al.
The Journal of Biological Chemistry, 286(29), 25922-25934 (2011)
Helena Maier et al.
The Journal of biological chemistry, 286(29), 25922-25934 (2011-06-02)
2-Hydroxylated fatty acid (HFA)-containing sphingolipids are abundant in mammalian skin and are believed to play a role in the formation of the epidermal barrier. Fatty acid 2-hydroxylase (FA2H), required for the synthesis of 2-hydroxylated sphingolipids in various organs, is highly
Fatty acid 2-Hydroxylation in mammalian sphingolipid biology
Hama H, et al.
Biochimica et Biophysica Acta - Molecular and Cell Biology of Lipids, 1801(4), 405-414 (2010)
Hiroko Hama
Biochimica et biophysica acta, 1801(4), 405-414 (2009-12-23)
2-Hydroxy fatty acids (hFA) are important components of a subset of mammalian sphingolipids. The presence of hFA in sphingolipids is best described in the nervous system, epidermis, and kidney. However, the literature also indicates that various hFA-sphingolipids are present in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service