860778C
Avanti
18:0 Dibromo MG (S-isomer)
Avanti Research™ - A Croda Brand, chloroform solution
Synonym(s):
1-(9,10-Dibromostearoyl)-sn-glycerol
Select a Size
About This Item
Recommended Products
Assay
>99% (TLC)
form
chloroform solution
mol wt
516.35 g/mol
packaging
pkg of 1 × 2.5 mL (860778C-25MG)
manufacturer/tradename
Avanti Research™ - A Croda Brand
concentration
10 mg/mL in chloroform (860778C-25MG)
shipped in
dry ice
storage temp.
−20°C
SMILES string
OC[C@@](O)([H])COC(CCCCCCCC(Br)C(Br)CCCCCCCC)=O
InChI
1S/C21H40Br2O4/c1-2-3-4-5-7-10-13-19(22)20(23)14-11-8-6-9-12-15-21(26)27-17-18(25)16-24/h18-20,24-25H,2-17H2,1H3/t18-,19?,20?/m0/s1
InChI key
XXBCOZLVGAZRSV-HDYDNRTBSA-N
Packaging
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service