Skip to Content
Merck
All Photos(1)

Key Documents

860682P

Avanti

Lyso GB3-d7 (synthetic)

powder

Synonym(s):

Globotriaosylsphingosine-d7

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$443.00

$443.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$443.00

About This Item

Empirical Formula (Hill Notation):
C36H60D7NO17
CAS Number:
Molecular Weight:
792.96
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.12

$443.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99% (TLC)

form

powder

packaging

package of 1 × 100 μg (860682P-100UG)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@H]1[C@H](OC[C@H](N)[C@H](O)/C=C/CCCCCCCCCCC([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])O[C@H](CO)[C@@H](O[C@@H]2O[C@H](CO)[C@@H]([C@H](O)[C@H]2O)O[C@H]3O[C@H](CO)[C@@H]([C@H](O)[C@H]3O)O)[C@@H]1O

InChI key

GRGNVOCPFLXGDQ-IZKFDWHNSA-N

General description

Globotriaosylsphingosine-d7 (Lyso GB3-d7) is a deuterated derivative of globotriaosylsphingosine (Lyso-GB3). Lyso GB3 is a bioactive glycolipid circulating in the blood.[1] It is a deacylated form of globotriaosylceramide.[2]

Biochem/physiol Actions

Globotriaosylsphingosine (Lyso GB3) accumulates to a greater extent in fabry disease.[2]

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (860682P-100UG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

The Fabry disease-associated lipid Lyso-Gb3 enhances voltage-gated calcium currents in sensory neurons and causes pain
Choi L, et al.
Neuroscience Letters, 594, 163-168 (2015)
Lyso-Gb3 activates Notch1 in human podocytes
Sanchez N, et al.
Human Molecular Genetics, 24(20), 5720-5732 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service