Skip to Content
Merck
All Photos(1)

Key Documents

860677P

Avanti

C18 Ceramide-d7 (d18:1-d7/18:0)

Avanti Research - A Croda Brand 860677P, powder

Synonym(s):

N-stearoyl-D-erythro-sphingosine (d7)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$1,130.00

$1,130.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$1,130.00

About This Item

Empirical Formula (Hill Notation):
C36H64D7NO3
CAS Number:
Molecular Weight:
573.00
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.25

$1,130.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 mg (860677P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860677P

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@]([H])(NC(CCCCCCCCCCCCCCCCC)=O)[C@@](O)([H])/C=C/CCCCCCCCCCC(C(C([2H])([2H])[2H])([2H])[2H])([2H])[2H]

InChI

1S/C36H71NO3/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-36(40)37-34(33-38)35(39)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2/h29,31,34-35,38-39H,3-28,30,32-33H2,1-2H3,(H,37,40)/b31-29+/t34-,35+/m0/s1/i2D3,4D2,6D2

InChI key

VODZWWMEJITOND-MUVBWDEFSA-N

General description

C18 Ceramide (d18:1/18:0) is a synthetic ceramide containing sphingosine with stearic acid. C18 Ceramide-d7 (d18:1-d7/18:0) is a deuterated derivative of ceramide d18:1/18:0.

Application

C18 Ceramide-d7 (d18:1-d7/18:0) has been used as a standard in liquid chromatography–mass spectrometry (LC-MS) for quantitative analysis of C18 ceramide in skeletal muscle.[1] It has also been used as a standard to measure C35–C39 ceramides in zebrafish brain and human embryonic kidney cell line (HEK293) by thermo q-exactive plus quadrupole-orbitrap mass spectrometery.[2]

Biochem/physiol Actions

C18 ceramide is involved in cell growth inhibition.[3] Determining the levels of the ceramides (Cer) d18:1/16:0, Cer d18:1/18:0, Cer d18:1/24:0, and Cer d18:1/24:1 and ratios in human plasma enables the prognosis of fatal outcome of coronary artery disease. Stable isotope form of C18 ceramide d18:1/18:0 supports a newly described validated liquid chromatography-tandem mass spectrometry methodology for their clinical-scaled measurement.[4]

Packaging

5 mL Amber Glass Screw Cap Vial (860677P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takhar Kasumov et al.
Analytical biochemistry, 401(1), 154-161 (2010-02-25)
We present an optimized and validated liquid chromatography electrospray ionization tandem mass spectrometry (LC-ESI-MS/MS) method for the simultaneous measurement of concentrations of different ceramide species in biological samples. The method of analysis of tissue samples is based on Bligh and
Tejia Zhang et al.
Scientific reports, 9(1), 19939-19939 (2019-12-29)
Extensive characterisations of the zebrafish genome and proteome have established a foundation for the use of the zebrafish as a model organism; however, characterisation of the zebrafish lipidome has not been as comprehensive. In an effort to expand current knowledge
Anna Zalewska et al.
Biomolecules, 9(12) (2019-12-19)
This is the first study to investigate the relationship between ceramides, the mitochondrial respiratory system, oxidative stress, inflammation, and apoptosis in the submandibular gland mitochondria of mice with insulin resistance (IR). The experiment was conducted on 20 male C57BL/6 mice
Paul T Reidy et al.
The Journal of physiology, 596(21), 5217-5236 (2018-09-09)
Insulin sensitivity (as determined by a hyperinsulinaemic-euglyceamic clamp) decreased 15% after reduced activity. Despite not fully returning to baseline physical activity levels, insulin sensitivity unexpectedly, rebounded above that recorded before 2 weeks of reduced physical activity by 14% after the recovery
Hady Razak Hady et al.
Journal of clinical medicine, 8(12) (2019-12-18)
The liver plays a central role in the glucose and lipid metabolism. Studies performed on animal models have shown an important role of lipid accumulation in the induction of insulin resistance. We sought to explain whether in obese humans, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service