Skip to Content
Merck
All Photos(1)

Key Documents

860665P

Avanti

4E,14Z-Sphingadiene

Avanti Research - A Croda Brand 860665P, powder

Synonym(s):

(2S,3R,4E,14Z)-2-aminooctadec-4,14-diene-1,3-diol

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$369.00

$369.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$369.00

About This Item

Empirical Formula (Hill Notation):
C18H35NO2
CAS Number:
Molecular Weight:
297.48
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$369.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 mg (860665P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860665P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@@](N)([H])[C@]([H])(O)/C=C/CCCCCCCC/C=C\CCC

InChI

1S/C18H35NO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-18(21)17(19)16-20/h4-5,14-15,17-18,20-21H,2-3,6-13,16,19H2,1H3/b5-4-,15-14+

InChI key

KWDXKYNWAKMLKK-YQMRQDNGSA-N

General description

4E,14Z-Sphingadiene is a unique sphingoid base containing C18 chain, with trans double bond at C-4 and cis double bond between C14 and C15 with a bent structure.[1][2] It is found in the plasma, lymph, brain, kidney and lungs of humans and mice.[1] 4E,14Z-Sphingadiene or d-erythro-1,3-dihydroxy-2-amino-4-trans-14-cis-octadecadiene is the second largest long-chain base of human plasma sphingomyelins.[2]

Biochem/physiol Actions

Sphingadiene, a vital component of sphingolipids facilitates numerous processes such as apoptosis and cell signaling.[3] It inhibits Akt/Wnt signaling and acts as an anticancer agent in colon cancer. [3][4]

Packaging

5 mL Amber Glass Screw Cap Vial (860665P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biosynthesis of the anti-lipid-microdomain sphingoid base 4, 14-sphingadiene by the ceramide desaturase FADS3
Jojima K, et al.
Faseb Journal (2020)
O Renkonen et al.
Journal of lipid research, 10(6), 687-693 (1969-11-01)
The dienic long-chain base (sphingadienine) of human plasma sphingomyelins has been identified as d-erythro-1,3-dihydroxy-2-amino-4-trans-14-cis-octadecadiene. A similar sphingosine was also detected in plasma sphingomyelins of rat, rabbit, and cat. The key reaction in the structural studies was partial reduction of sphingadienine
Occurrence, structure elucidation, biosynthesis, functions and synthesis of sphingadienes
U Abeytunga T
Mini-Reviews in Organic Chemistry, 12(3), 282-292 (2015)
Structure of plasma sphingadienine
Renkonen O and Hirvisalo EL
Journal of Lipid Research, 10(6), 687-693 (1969)
Keisuke Jojima et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(2), 3318-3335 (2020-01-10)
Sphingolipids are multifunctional lipids. Among the sphingolipid-component sphingoid bases, 4,14-sphingadiene (SPD) is unique such that it has a cis double bond with a bent structure. Although SPD was discovered half a century ago, its tissue distribution, biosynthesis, and degradation remain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service